Skip to Content
Merck
All Photos(1)

Documents

B3148

Sigma-Aldrich

Anti-Goat/Sheep IgG−Biotin antibody, Mouse monoclonal

clone GT-34, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Goat/Sheep IgG, Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

secondary antibodies

clone

GT-34, monoclonal

form

buffered aqueous solution

species reactivity

sheep, goat, bovine

should not react with

rat, canine, mouse, rabbit, guinea pig, chicken, horse, pig, feline, human

technique(s)

direct ELISA: 1:5,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Goat IgG (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
The antibody reacts with an epitope located on the heavy chain of goat IgG1 and IgG2.
The product binds to all goat/sheep IgG1

Application

Monoclonal Anti-Goat/Sheep IgG-Biotin antibody produced in mouse has been used in:
  • enzyme linked immuno sorbent assay (ELISA)
  • immunoblotting
  • dot blotting
  • immunohistochemistry
  • immunocytochemistry

Monoclonal Anti-Goat/Sheep IgG-Biotin antibody produced in mouse may be used for ELISA at a minimum working dilution of 1:5000. It was used at a working dilution of 1:1000 to visualize collagen type IV in mouse digit sections.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin allows for the binding of various labels such as avidin or streptavidin.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Li et al.
Scandinavian journal of immunology, 57(5), 410-422 (2003-05-20)
Involution of the thymus and alterations in the development of thymocytes are the most prominent features of age-related immune senescence. We have carried out a comparative analysis of thymocyte and stroma in rapid thymic involution DBA/2 (D2) strain of mice
Dynamic changes after murine digit amputation: the MRL mouse digit shows waves of tissue remodeling, growth, and apoptosis
Gourevitch DL, et al.
Wound Repair and Regeneration, 17(3), 447 -4455 (2009)
Sofia Halin Bergström et al.
Scientific reports, 6, 31805-31805 (2016-08-24)
Accumulating data indicates that tumor-derived extracellular vesicles (EVs) are responsible for tumor-promoting effects. However, if tumor EVs also prepare the tumor-bearing organ for subsequent tumor growth, and if this effect is different in low and high malignant tumors is not
Miguel A Ortega et al.
Antioxidants (Basel, Switzerland), 11(4) (2022-04-24)
Pancreatic cancer is a malignancy of rising prevalence, especially in developed countries where dietary patterns and sedentariness favor its onset. This malady ranks seventh in cancer-related deaths in the world, although it is expected to rank second in the coming
Lara Sánchez-Trujillo et al.
Journal of personalized medicine, 13(6) (2023-06-28)
Chronic venous disease (CVD) is a common condition that affects the veins in the lower limbs, resulting in a variety of symptoms, such as swelling, pain, and varicose veins (VVs). The plenty hormonal, hemodynamic and mechanical changes occurred in pregnancy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service