Skip to Content
Merck
All Photos(2)

Key Documents

AV44167

Sigma-Aldrich

Anti-SLC19A1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CHMD, Anti-FOLT, Anti-IFC1, Anti-REFC, Anti-RFC1, Anti-Solute carrier family 19 (folate transporter), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC19A1(6573)

General description

Solute carrier family 19 (folate transporter), member 1 (SLC19A1, CHMD, FOLT, RFC1, IFC1) is a major carrier-mediated transporter in mammalian cells for the uptake of reduced folates and antifolate-based anticancer drugs such as methotrexate. RFC activity can be considered as a potential marker for predicting response to antifolate chemotherapy.

Specificity

Anti-SLC19A1 polyclonal antibody reacts with human solute carrier family 19 (folate transporter), member 1.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC19A1

Application

Anti-SLC19A1 polyclonal antibody is used to tag solute carrier family 19 (folate transporter), member 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 19 (folate transporter), member 19 in carrier-mediated reduced folate uptake.

Biochem/physiol Actions

Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. SLC19A1 plays a role in maintaining intracellular concentrations of folate.Transport of folate compounds into mammalian cells can occur via receptor-mediated (see MIM 136430) or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: MVPSSPAVEKQVPVEPGPDPELRSWRHLVCYLCFYGFMAQIRPGESFITP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Letter to the editor.
Gokce Gurler et al.
Neuropathology and applied neurobiology, 50(2), e12969-e12969 (2024-03-18)
Vishal Sangha et al.
Fluids and barriers of the CNS, 21(1), 67-67 (2024-08-28)
Folates are a family of B9 vitamins essential for normal growth and development in the central nervous system (CNS). Transport of folates is mediated by three major transport proteins: folate receptor alpha (FRα), proton-coupled folate transporter (PCFT), and reduced folate
Camille Alam et al.
Molecular pharmaceutics, 14(11), 3848-3858 (2017-09-09)
Folates are essential for brain development and function. Folate transport in mammalian tissues is mediated by three major folate transport systems, i.e., reduced folate carrier (RFC), proton-coupled folate transporter (PCFT), and folate receptor alpha (FRα), known to be regulated by
Julian C Gilmore et al.
EBioMedicine, 75, 103771-103771 (2021-12-27)
Due to the critical role of folates in neurodevelopment, it is important to understand potential interactions between anti-HIV drugs used during pregnancy, and folate delivery pathways in the placenta. This study investigates the effect of dolutegravir (DTG) exposure on the
Gokce Gurler et al.
Fluids and barriers of the CNS, 20(1), 47-47 (2023-06-17)
Reduced folate carrier 1 (RFC1; SLC19a1) is the main responsible transporter for the B9 family of vitamins named folates, which are essential for normal tissue growth and development. While folate deficiency resulted in retinal vasculopathy, the expression and the role of RFC1 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service