Skip to Content
Merck
All Photos(3)

Key Documents

AV32439

Sigma-Aldrich

Anti-OTX2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Orthodenticle homeobox 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

human, bovine, dog, guinea pig, horse, rabbit, sheep, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OTX2(5015)

General description

OTX2 is a homeodomain transcription factor that regulates organ development. Studies in mice have shown that Otx2 is involved in determining the fate of retinal photoreceptor cells. This transcription factor has also been implicated in pineal gland development.
Rabbit Anti-OTX2 antibody recognizes canine, chicken, human, mouse, rat, rabbit, zebrafish, and bovine OTX2.

Immunogen

Synthetic peptide directed towards the N terminal region of human OTX2

Application

Rabbit Anti-OTX2 antibody can be used for western blot applications at a concentration of 1.0-10.0μg/ml. It can also be used for IHC applications at 4-8μg/ml.

Biochem/physiol Actions

Homeobox protein OTX2 is a member of the bicoid sub-family of homeodomain-containing transcription factors. This protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper forebrain development. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants may exist, but their full length sequences have not been determined. This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper forebrain development. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants may exist, but their full length sequences have not been determined.

Sequence

Synthetic peptide located within the following region: PESRVQVWFKNRRAKCRQQQQQQQNGGQNKVRPAKKKTSPAREVSSESGT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiro Nishida et al.
Nature neuroscience, 6(12), 1255-1263 (2003-11-20)
Understanding the molecular mechanisms by which distinct cell fate is determined during organogenesis is a central issue in development and disease. Here, using conditional gene ablation in mice, we show that the transcription factor Otx2 is essential for retinal photoreceptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service