Recommended Products
antibody form
purified immunoglobulin
species reactivity
human (IgG)
shipped in
wet ice
storage temp.
2-8°C
General description
A Protein Conformational Array (PCA) in a sandwich ELISA format where the plate is coated with a panel of antibodies raised against peptides derived from the full length protein sequence of Ustekinumab. Taken individually, each of these antibodies is strongly antigenic to the peptide sequence that was used in its production. However, when these peptides are incorporated into a full length correctly folded protein, the antigenicity of many of them is masked by the three dimensional structure of the protein and only a limited number of the antibodies respond. The result is a histogram which can be likened to a ‘fingerprint′ for correctly folded Ustekinumab. For a Stelara Biosimilar, if the protein is correctly folded and glycosylated, the ‘fingerprint′ will match that of Ustekinumab. If it is not correctly folded, previously masked peptide sequences will be exposed and will be recognized by the antibody made to that exposed sequence. In this way, changes in the ‘fingerprint′ generated by the ELISA will point out differences between the Biosimilar and authentic Ustekinumab.
Application
For research use only. Not for use in diagnostic procedures.
Other Notes
ELISA kit contains 3 96-well microplates and all the components necessary to complete an ELISA test.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frontiers in pharmacology, 4, 103-103 (2013-08-24)
Antibody arrays were developed to probe a monoclonal antibody's three-dimensional structure (3-D structure). Peptides with overlapping regions were designed to cover the whole mAb light chain and heavy chain, respectively, and used to generate polyclonal antibodies after the conjugation of
Monoclonal antibody higher order structure analysis by high throughput protein conformational array.
mAbs, 10(3), 397-405 (2018-01-10)
The elucidation of antibody higher order structure (HOS) is critical in therapeutic antibody development. Since HOS determines the protein bioactivity and chemo-physical properties, this knowledge can help to ensure that the safety and efficacy attributes are not compromised. Protein conformational
Biosimilar mAb in-process Sample Higher Order Structure Analysis with Protein Conformational Array ELISA.
Pharmaceut. Res., 1-11 (2016)
mAbs, 6(5), 1163-1177 (2014-12-18)
Remsima (infliximab) was recently approved as the world's first biosimilar monoclonal antibody (mAb) in both the European Union and Korea. To achieve this, extensive physicochemical characterization of Remsima in relation to Remicade was conducted in order to demonstrate the highly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service