Skip to Content
Merck
All Photos(1)

Key Documents

A9776

Sigma-Aldrich

α-Actinin from chicken gizzard

~80% α-actinin basis (SDS-PAGE), ammonium sulfate suspension

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

chicken gizzard

Assay

~80% α-actinin basis (SDS-PAGE)

form

ammonium sulfate suspension

mol wt

100 kDa

storage temp.

2-8°C

Gene Information

General description

α-Actinin exists as a rod-shaped antiparallel dimer with two elongated subunits. The domain region contains an N-terminal actin-binding domain with tandem calponin homology domains, a central tandem 3-helix motifs, and EF-hand motifs at C-terminus. It belongs to the spectrin superfamily and is localized in actin structures.

Application

α-Actinin from chicken gizzard has been used:
  • as an antigen to coat plates for the capture of anti-actinin antibody using enzyme-linked immunosorbent assay (ELISA) from tumor cell line clones
  • in in vitro motility experiments to test its effect on actin filament movement
  • to coat cantilevers for strengthening fluorescently labeled actin filaments in force measurement studies

Biochem/physiol Actions

α-Actinin binds actin and has the ability to crosslink actin cytoskeleton. It also plays a key role in the structural maintenance of the Z-disk of striated muscle.

Packaging

Package size based on protein content

Physical form

Suspension in 2 M (NH4)2SO4 containing 20 mM Tris acetate, pH 7.6, 20 mM sodium chloride, 0.1 mM EDTA, 15 mM β-mercaptoethanol and 1 mM phenylmethylsulfonyl fluoride

Preparation Note

Alpha-actinin ammonium sulfate suspension should be mixed sufficiently and further diluted prior to use. Protein precipitate will be present in solution with high protein concentrations.

Prepared using a modification of the procedure of Neidel, J.E. and Cuatrecasas, P., Biochem. Biophys. Res. Commun., 91, 152 (1979).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Katzemich et al.
PLoS genetics, 9(3), e1003342-e1003342 (2013-03-19)
The Drosophila Alp/Enigma family protein Zasp52 localizes to myotendinous junctions and Z-discs. It is required for terminal muscle differentiation and muscle attachment. Its vertebrate ortholog ZASP/Cypher also localizes to Z-discs, interacts with α-actinin through its PDZ domain, and is involved
F Rivero et al.
Journal of cell science, 112 ( Pt 16), 2737-2751 (1999-07-22)
The contribution of three actin cross-linking proteins, alpha-actinin (alphaA), gelation factor (ABP-120), and the 34 kDa actin-bundling protein to cellular functions has been studied in three single mutant (alphaA-, 120-, and 34-) and three double mutant (alphaA-/120-, 34-/alphaA-, 34-/120-) strains
Jun Liu et al.
Journal of molecular biology, 338(1), 115-125 (2004-03-31)
Cryoelectron microscopy was used to obtain a 3-D image at 2.0 nm resolution of 2-D arrays of smooth muscle alpha-actinin. The reconstruction reveals a well-resolved long central domain with 90 degrees of left-handed twist and near 2-fold symmetry. However, the
Flavius C Pascut et al.
Biochimica et biophysica acta, 1830(6), 3517-3524 (2013-02-14)
Online label-free monitoring of in-vitro differentiation of stem cells remains a major challenge in stem cell research. In this paper we report the use of Raman micro-spectroscopy (RMS) to measure time- and spatially-resolved molecular changes in intact embryoid bodies (EBs)
Norman Y Yao et al.
Physical review letters, 110(1), 018103-018103 (2013-02-07)
A hallmark of biopolymer networks is their sensitivity to stress, reflected by pronounced nonlinear elastic stiffening. Here, we demonstrate a distinct dynamical nonlinearity in biopolymer networks consisting of filamentous actin cross-linked by α-actinin-4. Applied stress delays the onset of relaxation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service