Skip to Content
Merck
All Photos(1)

Key Documents

A2522

Sigma-Aldrich

Actin from rabbit muscle

≥85% (SDS-PAGE), lyophilized powder

Synonym(s):

Alpha-actin 1, G-Actin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rabbit

Quality Level

Assay

≥85% (SDS-PAGE)

form

lyophilized powder

mol wt

43 kDa

composition

Protein, ≥80% biuret

technique(s)

immunoblotting: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

rabbit ... LOC(100342017)

Looking for similar products? Visit Product Comparison Guide

General description

Actin is the abundant protein found in many cells which accounts for 10% or more of total protein. Most of the eukaryotic cells shows the presence of actin protein. Outer and inner domains are the two important domains of actin.

Application

Actin from rabbit muscle has been used:
  • to study its protective role on rabbit muscle lactate dehydrogenase from ascorbate inhibition
  • as standard in SDS gel and immunoblotting
  • in the protein pin array assays

Biochem/physiol Actions

Actin plays a key role in protein-protein interactions. It participates in cell motility, cell shape maintenance and polarity to the regulation of transcription.

Physical form

Lyophilized powder containing Tris, ATP and CaCl2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarrod A Call et al.
Journal of applied physiology (Bethesda, Md. : 1985), 105(3), 923-932 (2008-06-28)
Duchenne muscular dystrophy is characterized by the absence of dystrophin from muscle cells. Dystrophic muscle cells are susceptible to oxidative stress. We tested the hypothesis that 3 wk of endurance exercise starting at age 21 days in young male mdx
Desmin cytoskeletal modifications after a bout of eccentric exercise in the rat
Barash IA, et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 91-98 (2002)
Actin and Actin-Binding Proteins
Pollard TD, et al.
Cold Spring Harbor Perspectives in Biology, 8(8) (2016)
Pooneh Vaziri et al.
Genetics, 216(1), 177-190 (2020-08-06)
Myosin is an essential motor protein, which in muscle is comprised of two molecules each of myosin heavy-chain (MHC), the essential or alkali myosin light-chain 1 (MLC1), and the regulatory myosin light-chain 2 (MLC2). It has been shown previously that MLC2
J A Henderson et al.
Endocrine, 33(3), 285-293 (2008-11-21)
To further understand the role of ovarian hormones in the function of the serotonin neural system, we investigated the effects of estradiol (E), progesterone (P), and raloxifene on 5HT 1A and 2C receptor protein expression in the dorsal raphe region

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service