A8849
α2-Antiplasmin from human plasma
lyophilized powder, ≥5 units/mg protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human plasma
Quality Level
form
lyophilized powder
specific activity
≥5 units/mg protein
packaging
vial of ≥0.5 mg (Protein based on E1%/280 = 6.7)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... SERPINF2(5345)
Looking for similar products? Visit Product Comparison Guide
General description
α2-Antiplasmin (α2AP) is a glycoprotein and a member of the serine protease inhibitor (serpin) family. The protein is mainly synthesized in the liver and found in the kidney and brain. The serpin peptidase inhibitor, clade F member 2 (SERPINF2) gene is mapped to human chromosome 17p13.3.
Application
α2-Antiplasmin from human plasma has been used as a protease inhibitor to treat human microvascular endothelial cells (HMEC-1) and demonstrate the role of factors responsible for endothelial cells detachment. It has also been used to study its effect on the release of proteolytic activity under reducing conditions in therapeutic human serum albumin-containing products with a new neoepitope endopeptidase immunoassay.
Biochem/physiol Actions
α2-Antiplasmin (α2AP) is an antifibrinolytic protein and an important inhibitor of plasmin. α2AP mediates clot formation by covalently cross-linking with the glutamine and the lysine residue of fibrin clot via activated factor XIII (FXIIIa). This prevents the clot from extreme degradation by plasmin. An inborn insufficiency of α2AP results in an autosomal recessive condition leading to a rare bleeding disorder. The severity of the bleeding condition is defined by the individual′s genetic variation. Studies show lower levels of α2AP associated with disseminated intravascular coagulation (DIC) and thrombolytic therapy. Elevated levels of α2AP have been reported in a patient with ischemic stroke.
Unit Definition
One unit will reduce the activity of two units of plasmin by 50% at 37 °C, pH 7.2, using Val-Leu-Lys-p-nitroanilide.
Physical form
Lyophilized powder containing Bis-tris buffer and NaCl.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yannève Rolland et al.
Biochimica et biophysica acta, 1763(4), 393-401 (2006-05-23)
Tissue plasminogen activator (t-PA) is an extracellular serine protease that converts the proenzyme plasminogen into the broad-spectrum substrate serine protease, plasmin. Plasmin, one of the most potent pro-angiogenic factors, is a key element in fibrinolysis, cell migration, tissue remodeling and
Nikoletta Pechlivani et al.
International journal of molecular sciences, 22(22) (2021-11-28)
Thrombus formation remains a major cause of morbidity and mortality worldwide. Current antiplatelet and anticoagulant therapies have been effective at reducing vascular events, but at the expense of increased bleeding risk. Targeting proteins that interact with fibrinogen and which are
Russell G A Jones et al.
Journal of pharmaceutical and biomedical analysis, 54(1), 74-80 (2010-09-14)
Botulinum type A toxin (BoNT/A) is defined by its specific endopeptidase cleavage of SNAP25 between Gln(197) and Arg(198) under reducing conditions. The neurotoxin is widely used for therapeutic or cosmetic purposes, but should not contain other toxin serotypes or unwanted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service