Skip to Content
Merck
All Photos(2)

Key Documents

A7165

Sigma-Aldrich

1,5-Anhydro-D-sorbitol

crystalline, ≥98.0% (TLC)

Synonym(s):

1,5-Anhydro-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (TLC)

form

crystalline

color

white

solubility

water: 49.00-51.00 mg/mL, clear, colorless

SMILES string

OCC1OCC(O)C(O)C1O

InChI

1S/C6H12O5/c7-1-4-6(10)5(9)3(8)2-11-4/h3-10H,1-2H2

InChI key

MPCAJMNYNOGXPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,5-Anhydro-D-sorbitol, a cyclic polyol, is a metabolic product of 1,5 anhydro-D-fructose. 1,5-Anhydro-D-sorbitol is used as a reference in analytical procedures that assess the metabolism of 1,5 anhydro-D-fructose, a food supplement with potential benefits as an antioxidant, antimicrobial, and antidiabetic agent.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej S Januszewski et al.
Diabetes research and clinical practice, 95(1), e17-e19 (2011-10-26)
An assay of plasma 1,5-anhydroglucitol was evaluated. Assay CVs, effects of four plasma freeze-thaw cycles, glucose up to 80 mmol/L and triglycerides up to 20 mmol/L were acceptable. 1,5-anhydroglucitol levels were significantly lower in diabetic vs. non-diabetic subjects and correlated
Sanjeev N Mehta et al.
Pediatric diabetes, 13(3), 278-284 (2011-11-09)
Serum 1,5-anhydroglucitol (1,5-AG) is a marker of hyperglycemic excursions in adults with diabetes and hemoglobin A1c (HbA1c) < 8%. We compared 1,5-AG levels among youth and young adults with and without type 1 diabetes (T1D) and investigated the utility of
Ai Ogawa et al.
Chemical & pharmaceutical bulletin, 59(5), 672-675 (2011-05-03)
A new compound, pycnalin (1), together with four known compounds, ginnalins A (2), B (3), C (4), and 3,6-di-O-galloyl-1,5-anhydro-D-glucitol (3,6-di-GAG) (5), were isolated from Acer pycnanthum. The structure of 1 was determined on the basis of 2D-NMR spectral data and
Rimesh Pal et al.
Diabetic medicine : a journal of the British Diabetic Association, 38(3), e14515-e14515 (2021-01-10)
To summarize all relevant randomized controlled trials (RCTs) and provide precise effect estimates of glycaemic efficacy/safety of faster-acting insulin aspart administered by injection as compared to insulin aspart in people with diabetes mellitus. PubMed/Cochrane Library were systematically searched till October
Stephen P Juraschek et al.
Diabetes care, 35(11), 2265-2270 (2012-08-10)
Fructosamine, glycated albumin, and 1,5-anhydroglucitol (1,5-AG) are of interest for monitoring short-term glycemic control in patients with diabetes; however, their associations with diabetes risk are uncharacterized. We used Cox proportional hazards models to examine the associations of fructosamine, glycated albumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service