Skip to Content
Merck
All Photos(1)

Documents

A6888

Sigma-Aldrich

Adenosine 5′-triphosphate–Agarose

aqueous glycerol suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

aqueous glycerol suspension

extent of labeling

≥1 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

ribose hydroxyls

matrix spacer

11 atoms (adipic acid dihydrazide)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate Agarose (5′-ATP agarose) has been used in affinity chromatography to purify uridine kinase from Ehrlich ascites tumor cells.

Physical form

Suspension in 50% glycerol containing 0.25 M NaCl

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Stitt
The Journal of biological chemistry, 263(23), 11130-11137 (1988-08-15)
We have determined that 3 mol of ATP or other adenine nucleotide can bind to Escherichia coli transcription termination protein rho, in the presence or absence of the RNA cofactor that is required for activation of rho's ATPase activity. Isotope
S Ogg et al.
The Journal of biological chemistry, 269(48), 30461-30469 (1994-12-02)
Human Cdc25C is a protein phosphatase that dephosphorylates and activates Cdc2-cyclin B to trigger entry into mitosis. Cdc25C is itself regulated by phosphorylation. In asynchronously growing HeLa cells, we have determined that serine 216 is the major site of Cdc25C
V Nagaraja et al.
Journal of molecular biology, 182(4), 579-587 (1985-04-20)
We have purified the type I restriction enzymes SB and SP from Salmonella typhimurium and S. potsdam, respectively, and determined the DNA sequences that they recognize. These sequences resemble those previously determined for the type I enzymes, EcoB, EcoK and
B Suri et al.
The EMBO journal, 3(3), 575-579 (1984-03-01)
The EcoA restriction enzyme from Escherichia coli 15T- has been isolated. It proves to be an unusual enzyme, clearly related functionally to the classical type I restriction enzymes. The basic enzyme is a two subunit modification methylase. Another protein species
E Scherzinger et al.
The Journal of biological chemistry, 272(48), 30228-30236 (1997-12-31)
The RepA protein of the mobilizable broad host range plasmid RSF1010 has a key function in its replication. RepA is one of the smallest known helicases. The protein forms a homohexamer of 29,896-Da subunits. A variety of methods were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service