Skip to Content
Merck
All Photos(4)

Key Documents

A3903

Sigma-Aldrich

1-Aminocyclopropanecarboxylic acid

≥98% (TLC), powder, NMDA agonist

Synonym(s):

ACC, ACPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
Beilstein:
2076413
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Product Name

1-Aminocyclopropanecarboxylic acid, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.
Keep in a dry place.)

color

white

mp

229-231 °C (lit.)

solubility

water: 49.00-51.00 mg/mL, clear, colorless

SMILES string

NC1(CC1)C(O)=O

InChI

1S/C4H7NO2/c5-4(1-2-4)3(6)7/h1-2,5H2,(H,6,7)

InChI key

PAJPWUMXBYXFCZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

1-Aminocyclopropanecarboxylic acid (ACC) is a non-proteinogenic amino acid and is a member of 1-aminocycloalkanecarboxylic acids.

Application

1-Aminocyclopropanecarboxylic acid (ACC) has been used as a substrate for determining ACC deaminase activity to study the plant growth-promoting characteristics of endophytic isolates obtained from Pteris vittata roots, cucurbit seeds. It has also been used as an ethylene precursor to study the effect of high ethylene concentration on lateral root density (LRD) in sorghum brown midrib (bmr) mutants.

Biochem/physiol Actions

1-Aminocyclopropanecarboxylic acid (ACC) is a partial agonist acting at the N-methyl-D-aspartate (NMDA) receptor glycine modulatory site. ACC serves as a bio-synthetic precursor for the plant hormone ethylene. Therefore, it also plays a vital role in plant development. ACC exhibits anticonvulsant, anxiolytic, anti-addictive, neuroprotective and antidepressant-like effects in rats and mice. ACC does not affect memory or learning processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Sarita Tiwari et al.
Journal of environmental management, 180, 359-365 (2016-06-04)
Mitigation of arsenic (As) pollution is a topical environmental issue of high R&D priority. The present investigation was carried out to isolate As resistant endophytes from the roots of Indian ecotype Pteris vittata and characterize their As transformation and tolerance
Joanna K Polko et al.
Frontiers in plant science, 10, 1602-1602 (2020-01-11)
1-Aminocyclopropane 1-carboxylic acid (ACC) is the direct precursor of the plant hormone ethylene. ACC is synthesized from S-adenosyl-L-methionine (SAM) by ACC synthases (ACSs) and subsequently oxidized to ethylene by ACC oxidases (ACOs). Exogenous ACC application has been used as a
Y A Kolesnikov et al.
Life sciences, 55(18), 1393-1398 (1994-01-01)
1-Aminocyclopropane carboxylic acid (ACPC), a partial agonist of the glycine site on the NMDA receptor, prevents tolerance to the mu opioid morphine and the delta ligand [D-Pen2,D-Pen5]enkephalin (DPDPE) when co-administered with the opioid. In contrast, ACPC does not significantly influence
Valentin Marquis et al.
Plant, cell & environment, 43(6), 1558-1570 (2020-03-13)
Jasmonate synthesis and signalling are essential for plant defense upregulation upon herbivore or microbial attacks. Stress-induced accumulation of jasmonoyl-isoleucine (JA-Ile), the bioactive hormonal form triggering transcriptional changes, is dynamic and transient because of the existence of potent removal mechanisms. Two
Lorenzo Poncini et al.
PloS one, 12(10), e0185808-e0185808 (2017-10-04)
Plants interpret their immediate environment through perception of small molecules. Microbe-associated molecular patterns (MAMPs) such as flagellin and chitin are likely to be more abundant in the rhizosphere than plant-derived damage-associated molecular patterns (DAMPs). We investigated how the Arabidopsis thaliana

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service