Skip to Content
Merck
All Photos(2)

Key Documents

A3098

ART® self-sealing barrier pipette tips

ART 200, volume range 1-200 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless

sterility

sterile

feature

barrier

packaging

case of 5 packs
pack of 960 ea (Racked)

manufacturer/tradename

Molecular BioProducts 2069

technique(s)

PCR: suitable

maximum volume

200 μL

volume range

1-200 μL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Achieve consistent and precise performance in your applications with the reliable ART® self-sealing barrier pipette tips. These tips feature a sealing mechanism that effectively traps potential contaminants inside the barrier, ensuring sample integrity. It is designed to support a wide range of pipetting tasks. The ART barrier pipette tips provide convenience and versatility.
Generous filter placement and compatibility with many 200 μL pipetters

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).
ART® self-sealing barrier pipette tips suitable for loading gels, including acrylamide or agarose gels. It is also ideal for handling genomic DNA, fragile cell lines and other viscous materials.

Features and Benefits

  • Certified sterile tips that are free from RNase, DNase, and pyrogen contamination.
  • Options available for both sterile and nonsterile tips, with various packaging choices.
  • Manufactured in accordance with the highest quality standards of the industry.
  • Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel.
  • Solvent safe and is suitable for use with acids, bases, and aggressive organic solvents.

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit mLINE 20-200 μl
Biohit Variable 50-200 μl
Biohit Proline-Variable 50-200 μl
Capp Aero 200 μl
Corning 4963 20-200 μl
Eppendorf Research/Reference 20-200 μl
Eppendorf Research Pro 4860 20-300 μl
Eppendorf Response 4850 20-500 μl
Eppendorf Titerman 4908 30-300 μl
Finnpipette 4027/4500 40-200 μl
Mettler-Toledo VoluMate 20-200 μl
MLA Digital Pipette 50-200 μl
Nichiryo BenchMate 20-200 μl
Pipetman P-200 20-200 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Rainin Pipet Plus 20-200 μl
Rainin Pipetman 20-200 μl
Transferpette 20-200 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service