Skip to Content
Merck
All Photos(1)

Key Documents

A2723

ART® self-sealing barrier pipette tips

ART 20E, volume range 0.5-10 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
10 μL
volume range:
0.5-10 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
case of 5 packs
pack of 960 ea (Racked)
manufacturer/tradename:
Molecular BioProducts 2149E

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

case of 5 packs
pack of 960 ea (Racked)

manufacturer/tradename

Molecular BioProducts 2149E

technique(s)

PCR: suitable

maximum volume

10 μL

volume range

0.5-10 μL

Looking for similar products? Visit Product Comparison Guide

General description

An ultra-micro tip designed for Eppendorf and Nichiryo/BenchMate pipetters.

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Eppendorf 4810 .5-10 μl
Eppendorf Research/Reference .5-10 μl
Eppendorf Research Pro .5-10 μl
Eppendorf Response 4850 .5-10 μl
Eppendorf Titerman 4908 .5-10 μl
Mettler-Toledo VoluMate .5-10 μl
Nichiryo/BenchMate .5-10 μl
Socorex/Wheaton Calibra/822 .2-2 μl/1-10 μl
Transferpette Single/Multi-Channel .5-10 μl
Transferpette Multi-Channel (Limited Draw) 2-20 μl
Transferpette Ultra-Micro 2-20 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service