A2380
2-Aminopurine nitrate salt
≥99%
Synonym(s):
2-AP nitrate salt
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Empirical Formula (Hill Notation):
C5H5N5 · HNO3
CAS Number:
Molecular Weight:
198.14
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
biological source
synthetic
Assay
≥99%
form
powder
solubility
water: 50 mg/mL, clear, faintly to light yellow
SMILES string
O[N+]([O-])=O.Nc1ncc2[nH]cnc2n1
InChI
1S/C5H5N5.HNO3/c6-5-7-1-3-4(10-5)9-2-8-3;2-1(3)4/h1-2H,(H3,6,7,8,9,10);(H,2,3,4)
InChI key
PGDNOZBGABWZLD-UHFFFAOYSA-N
Application
2-Aminopurine (2-AP) is used to specifically inhibit double-stranded RNA-dependent protein kinase, protein kinase R (PKR).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yasumi Endoh et al.
Journal of immunology (Baltimore, Md. : 1950), 182(4), 2258-2268 (2009-02-10)
The S100 calcium-binding proteins S100A8 and S100A9 are elevated systemically in patients with viral infections. The S100A8-S100A9 complex facilitated viral replication in human CD4(+) T lymphocytes latently infected with HIV-1- and S100A8-induced HIV-1 transcriptional activity. Mechanisms inducing the S100 genes
Junpei Teramachi et al.
Experimental cell research, 316(19), 3254-3262 (2010-08-24)
Double-stranded RNA-dependent protein kinase (PKR) plays a critical role in antiviral defence of the host cells. PKR is also involved in cell cycle progression, cell proliferation, cell differentiation, tumorigenesis, and apoptosis. We previously reported that PKR is required for differentiation
Renata M S Pereira et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(2), 617-626 (2009-10-09)
The evolution of Leishmania infection depends on the balance between microbicidal and suppressor macrophage functions. Double-stranded RNA (dsRNA)-activated protein kinase R (PKR), a classic antiviral protein, is able to regulate a number of signaling pathways and macrophage functions. We investigated
Michael E Lindquist et al.
Virology, 413(1), 103-110 (2011-03-08)
We performed experiments to determine the effect of PKR activation on respiratory syncytial virus (RSV) replication. We first determined that RSV infection activates PKR which induces the phosphorylation of eIF2α, resulting in the formation of host stress granules. We used
Chen-Hsuan Wang et al.
Brain research bulletin, 79(3-4), 169-176 (2009-05-12)
Molecules involved in self-protection of neurons against glucose/oxygen/serum deprivation (GOSD) were investigated. Trypan blue dye exclusion assay, Western blotting, ELISA, cytokine antibody array and chemical blocking assay were applied in the study. Results showed that early induction (at 6h of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service