Skip to Content
Merck
All Photos(1)

Key Documents

94259

Sigma-Aldrich

(±)-Mevalonic acid 5-pyrophosphate tetralithium salt

≥80% (qNMR)

Synonym(s):

(±)-MVAPP, (±)-Mevalonic acid 5-diphosphate tetralithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10Li4O10P2
CAS Number:
Molecular Weight:
331.85
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥80% (qNMR)

form

powder

color

white

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].CC(O)(CCOP([O-])(=O)OP([O-])([O-])=O)CC([O-])=O

InChI

1S/C6H14O10P2.4Li/c1-6(9,4-5(7)8)2-3-15-18(13,14)16-17(10,11)12;;;;/h9H,2-4H2,1H3,(H,7,8)(H,13,14)(H2,10,11,12);;;;/q;4*+1/p-4

InChI key

GFIXUUFXTAPBAD-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.
Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase. Mevalonic acid 5-pyrophosphate was shown to elevate ubiquinone levels in rat liver tissues.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Lombard et al.
Molecular biology and evolution, 28(1), 87-99 (2010-07-24)
Isoprenoids are a very diverse family of organic compounds widespread in the three domains of life. Although they are produced from the condensation of the same precursors in all organisms (isopentenyl pyrophosphate and dimethylallyl diphosphate), the evolutionary origin of their
Luke Grundy et al.
Pain, 160(4), 793-804 (2018-12-12)
Primary afferent neurons transduce distension of the bladder wall into action potentials that are relayed into the spinal cord and brain, where autonomic reflexes necessary for maintaining continence are coordinated with pathways involved in sensation. However, the relationship between spinal
Muthusubramanian Venkateshwaran et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(31), 9781-9786 (2015-07-23)
Rhizobia and arbuscular mycorrhizal fungi produce signals that are perceived by host legume receptors at the plasma membrane and trigger sustained oscillations of the nuclear and perinuclear Ca(2+) concentration (Ca(2+) spiking), which in turn leads to gene expression and downstream
D McCaskill et al.
Analytical biochemistry, 215(1), 142-149 (1993-11-15)
Procedures for the isolation and analysis of all 11 intermediates of the mevalonic acid pathway from acetyl-CoA through geranylgeranyl pyrophosphate were developed. Both acid-labile and base-labile metabolites are simultaneously extracted with good recoveries into 7 M urea at neutral pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service