Skip to Content
Merck
All Photos(1)

Key Documents

M4667

Sigma-Aldrich

(±)-Mevalonolactone

~97% (titration)

Synonym(s):

(±)-β-Hydroxy-β-methyl-δ-valerolactone, (±)-3-Hydroxy-3-methyl δ-valerolactone, DL-Mevalolactone, DL-Mevalonic acid lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
80960
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.25

Assay

~97% (titration)

storage temp.

−20°C

SMILES string

CC1(O)CCOC(=O)C1

InChI

1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3

InChI key

JYVXNLLUYHCIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alkaline hydrolysis of mevalonolactone gives mevalonate. Mevalonate is a precursor of farnesyl and geranylgeranyl pyrophosphates. These pyrophosphates are required for protein prenylation.

Application

(±)-Mevalonolactone is used to:
  • study the effect of statin on the prenylation of Ras and Rho GTPases
  • analyse the isoprenoid biosynthesis pathways in Listeria monocytogenes
  • study the the effects of statins on proliferation and migration of HUVECs (HGF-induced human umbilical vein endothelial cells)

Other Notes

May also be liquid!

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of the isoprenoid biosynthesis pathways in Listeria monocytogenes reveals a role for the alternative 2-C-methyl-D-erythritol 4-phosphate pathway in murine infection.
Begley M
Infection and Immunity, 76, 5392-5401 (2008)
The effects of different types of statins on proliferation and migration of HGF-induced Human Umbilical Vein Endothelial Cells (HUVECs).
Burgazli KM
European Review for Medical and Pharmacological Sciences, 17, 2874-2883 (2013)
Coenzyme Q 10 production in recombinant Escherichia coli strains engineered with a heterologous decaprenyl diphosphate synthase gene and foreign mevalonate pathway.
Zahiri HS
Metabolic engineering, 8, 406-416 (2006)
Svena M Lüdke et al.
Journal of lipid research, 49(12), 2620-2626 (2008-08-02)
The aim of this study was to investigate the possible existence and magnitude of stable carbon isotope discrimination by human 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR). The catalytic portion of HMGR was expressed and purified. The reaction product mevalonate was lactonized and
Mareike Bongers et al.
eLife, 9 (2020-03-13)
Volatile isoprenoids produced by plants are emitted in vast quantities into the atmosphere, with substantial effects on global carbon cycling. Yet, the molecular mechanisms regulating the balance between volatile and non-volatile isoprenoid production remain unknown. Isoprenoids are synthesised via sequential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service