Skip to Content
Merck
All Photos(1)

Documents

78331

Sigma-Aldrich

FITC-Dextran sulfate sodium salt

average mol wt 40,000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

biological source

synthetic

Quality Level

conjugate

FITC conjugate

form

powder

mol wt

average mol wt 40,000

composition

sulfur content, 16-20%

extent of labeling

0.001-0.020 mol FITC per mol glucose (substitution)

loss

≤10% loss on drying

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Howard et al.
Journal of cell science, 132(7) (2019-03-07)
Basement membranes are an ancient form of animal extracellular matrix. As important structural and functional components of tissues, basement membranes are subject to environmental damage and must be repaired while maintaining functions. Little is known about how basement membranes get
Jeremy J Cottrell et al.
Animals : an open access journal from MDPI, 10(4) (2020-04-25)
With increases in the frequency, intensity and duration of heat waves forecast plus expansion of tropical agriculture, heat stress (HS) is both a current and an emerging problem. As cinnamon has been shown to increase insulin sensitivity, which is part

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service