Skip to Content
Merck
All Photos(1)

Documents

53319

Sigma-Aldrich

L-Histidine

≥99.5% (NT), BioUltra

Synonym(s):

(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Histidine, BioUltra, ≥99.5% (NT)

product line

BioUltra

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D −39±1°, c = 2% in H2O

impurities

insoluble matter, passes filter test
≤0.5% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 110 °C

color

white

pH

7.0-8.0 (25 °C, 0.1 M in H2O)

mp

282 °C (dec.) (lit.)
~285 °C (dec.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

application(s)

detection

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Application

L-Histidine has been used for characterization of ZnO nanorod in an attempt to develop a technique for the ultra-low level detection of l-histidine.

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.
L-Histidine is involved in the one-carbon unit metabolism. It is associated with protein methylation. L-Histidine is a part of hemoglobin structure and function. L-Histidine is a component of dipeptides with antioxidative property. Histidine serves as a precursor for the formation of histamine, which is associated with allergic responses. Urocanic acid, an immune response modulator in skin is also biosynthesised from histidine.

Other Notes

Amino acid spacing in isotachophoresis on polyacrylamide gels - a critical evaluation; Prevents circular DNA strand cleavage in a xanthine-xanthine oxidase system; Growth requirement of various microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-Low Level Detection of L-Histidine Using Solution-Processed ZnO Nanorod on Flexible Substrate.
Sasmal M
IEEE Transactions on Nanobioscience, 14(6), 634-640 (2015)
K Brawn et al.
Acta physiologica Scandinavica. Supplementum, 492, 9-18 (1980-01-01)
An enzymic flux of O2- and H2O2 caused strand breaks in the supercoiled covalently closed circular Col El plasmid. Subnanomolar levels of superoxide levels of superoxide dismutase or of catalase prevented this attack on DNA, signifying that both O2- and
CRC Handbook of Microbiology, 4, 1-1 (1974)
Amino acid spacing in isotachophoresi; on polyacrylamide gel: a critical evaluation.
N Y Nguyen et al.
Analytical biochemistry, 94(1), 202-210 (1979-04-01)
H M Golder et al.
Journal of dairy science, 97(8), 5131-5150 (2014-06-03)
Ruminal bacterial community composition (BCC) and its associations with ruminal fermentation measures were studied in dairy heifers challenged with combinations of grain, fructose, and histidine in a partial factorial study. Holstein-Friesian heifers (n=30) were randomly allocated to 5 triticale grain-based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service