H7750
DL-Histidine
≥99% (TLC), suitable for ligand binding assays
Synonym(s):
(±)-2-Amino-3-(4-imidazolyl)propionic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
7800
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
DL-Histidine, ≥99% (TLC)
Assay
≥99% (TLC)
form
powder
technique(s)
ligand binding assay: suitable
mp
273 °C (dec.) (lit.)
solubility
0.5 M HCl: soluble
application(s)
peptide synthesis
SMILES string
NC(Cc1c[nH]cn1)C(O)=O
InChI
1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)
InChI key
HNDVDQJCIGZPNO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
DL-Histidine has been used to study the fouling behaviour of polyethersulfone ultrafiltration membranes made with different PVP (polyvinylpyrrolidone).
DL-Histidine is the mixed isomer of D- and L-histidine. DL-Histidine may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis.
Biochem/physiol Actions
L-Histidine is involved in the one-carbon unit metabolism. It is associated with protein methylation. L-Histidine is a part of hemoglobin structure and function. L-Histidine is a component of dipeptides with antioxidative property. Histidine serves as a precursor for the formation of histamine, which is associated with allergic responses. Urocanic acid, an immune response modulator in skin is also biosynthesised from histidine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fouling behaviour of polyethersulfone UF membranes made with different PVP
J.Marchese
Journal of Membrane Science , 211, 1-11 (2009)
Worapan Pormsila et al.
Electrophoresis, 32(8), 884-889 (2011-03-12)
The quantification of plasma lactate and evaluation of the lactate threshold by CE with capacitively coupled contactless conductivity is demonstrated. The only sample preparation needed was deproteinization with a ACN/methanol mixture. A solution of 10 mmol/L 2-morpholinoethanesulfonic acid monohydrate, 10
Tetsuya Chujo et al.
PloS one, 9(6), e98737-e98737 (2014-06-04)
WRKY transcription factors and mitogen-activated protein kinase (MAPK) cascades have been shown to play pivotal roles in the regulation of plant defense responses. We previously reported that OsWRKY53-overexpressing rice plants showed enhanced resistance to the rice blast fungus. In this
Yoshikazu Tsukasaki et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(45), 16011-16016 (2014-10-31)
The cadherins Fat and Dachsous regulate cell polarity and proliferation via their heterophilic interactions at intercellular junctions. Their ectodomains are unusually large because of repetitive extracellular cadherin (EC) domains, which raises the question of how they fit in regular intercellular
Hai Zhou et al.
Nature communications, 5, 4884-4884 (2014-09-12)
Thermosensitive genic male-sterile (TGMS) lines, which are male-sterile at restrictive (high) temperatures but male-fertile at permissive (low) temperatures, have been widely used in breeding two-line hybrid rice (Oryza sativa L.). Here we find that mutation of thermosensitive genic male sterile
Chromatograms
application for HPLCapplication for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service