Skip to Content
Merck
All Photos(1)

Key Documents

5126

Sigma-Aldrich

CD274 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human

Quality Level

recombinant

expressed in E. coli

description

0.1 mg recombinant human CD274 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

technique(s)

cell culture | mammalian: suitable

accession no.

NP_054862.1

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CD274(29126)

Application

Coating a plate well (6 well plate) with this recombinant CD274 protein in a specific culture medium at 5-10 μg/well allows for use 1) as human T and B cells activation/differentiation studies or 2) as a potential biomarker protein for infectious diseases in vitro or 3) for auto-immuno disease diagnostic development.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1.Thaw CD274 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so the volume added covers the surface evenly (5-10 μg/well, 6 well plate).
2.Add appropriate amount of diluted material to culture surface.
3.Incubate at room temperature for approximately 1.5 hours.
4.Aspirate remaining material.
5.Rinse plates carefully with water and avoid scratching bottom surface of plates.
6.Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQG^GEFFTVTVPKDLYVVEYGSNMTIECKFPVEKQLDLAALIVYWEMEDKNIIQFVHGEEDLKVQHSSYRQRARLLKDQLSLGNAALQITDVKLQDAGVYRCMISYGGADYKRITVKVNAPYNKINQRILVVDPVTSEHELTCQAEGYPKAEVIWTSSDHQVLSGKTTTTNSKREEKLFNVTSTLRINTTTNEIFYCTFRRLDPEENHTAELVIPELPLAHPPNER

Preparation Note

The extracellular domain of recombinant human CD274 (19-238 aa, derived from BC074984) was constructed with codon optimization and expressed with a small T7-His-TEV cleavage site Tag (29aa) fusion at its N-terminal and expressed in E. coli as inclusion bodies. The final product was refolded using our unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoba Amarnath et al.
Science translational medicine, 3(111), 111ra120-111ra120 (2011-12-03)
Immune surveillance by T helper type 1 (T(H)1) cells is not only critical for the host response to tumors and infection, but also contributes to autoimmunity and graft-versus-host disease (GVHD) after transplantation. The inhibitory molecule programmed death ligand 1 (PDL1)
Daria Trabattoni et al.
Blood, 101(7), 2514-2520 (2002-12-07)
The ligation of programmed death-ligand 1 (B7-H1) to T cells results in the preferential production of interleukin 10 (IL-10). We investigated if B7-H1 would be up-regulated in HIV infection, a disease characterized by increased IL-10 production, by measuring B7-H1, B7-1
Yujia Cao et al.
Cancer research, 71(14), 4737-4741 (2011-07-07)
B7-H1 (CD274), a member of the B7 family of coinhibitory molecules, is often induced in human tumors and its expression is closely correlated with a poor prognosis or higher malignancy grade. Tumor-associated B7-H1 is implicated in mechanisms of immune escape.
Jun Yang et al.
Journal of immunology (Baltimore, Md. : 1950), 187(3), 1113-1119 (2011-06-24)
The programmed death ligand 1 (PDL1)/programmed death 1 (PD1) costimulatory pathway plays an important role in the inhibition of alloimmune responses as well as in the induction and maintenance of peripheral tolerance. It has been demonstrated recently that PDL1 also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service