44039
Atto 488 DPPE
suitable for fluorescence, ≥80% (TLC)
Synonym(s):
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine labeled with Atto 488
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥80% (TLC)
manufacturer/tradename
ATTO-TEC GmbH
λ
in chloroform/methanol (8:2)
UV absorption
λ: 505-511 nm Amax
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto 488 is a labeling dye with high molecular absorption (90,000) and quantum yield (0.80) as well as sufficient Stokes shift between excitation and emission maximum. It is optimized for excitation with an argon laser, and is characterized by high photostability.
Atto-Dye Labeled Phospholipids
Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head group. After incorporation of the phospholipid into a membrane the fluorophore is located at the water/lipid interface of the membrane. We currently provide 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), palmitoyl-sn-glycero-phosphoethanolamine (PPE), and 1,2-dimyristoyl-snglycero-3-phospho-ethanolamine (DMPE) labeled with Atto-dyes.
find more information here
Atto-Dye Labeled Phospholipids
Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head group. After incorporation of the phospholipid into a membrane the fluorophore is located at the water/lipid interface of the membrane. We currently provide 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), palmitoyl-sn-glycero-phosphoethanolamine (PPE), and 1,2-dimyristoyl-snglycero-3-phospho-ethanolamine (DMPE) labeled with Atto-dyes.
find more information here
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Breaking the Diffraction Barrier in Fluorescence Microscopy by Optical Shelving.
Physical Review Letters, 98(21) (2007)
Instant Live-Cell Super-Resolution Imaging of Cellular Structures by Nanoinjection of Fluorescent Probes.
Nano Letters, 15(2), 1374-1381 (2015)
The changing point-spread function. Single-molecule-based super-resolution imaging.
Histochemistry and Cell Biology, 141(6), 577-585 (2014)
Dynamic Superresolution Imaging of Endogenous Proteins on Living Cells at Ultra-High Density.
Biophysical Journal, 99(4), 1303-1310 (2010)
Dual color localization microscopy of cellular nanostructures.
Biotechnology Journal, 4(6), 927-938 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service