Skip to Content
Merck
All Photos(1)

Key Documents

375M-9

Sigma-Aldrich

HGAL (MRQ-49) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-49, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (375M-94)
vial of 0.5 mL concentrate (375M-95)
bottle of 1.0 mL predilute (375M-97)
vial of 1.0 mL concentrate (375M-96)
bottle of 7.0 mL predilute (375M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2aκ

control

lymph node, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GCSAM(257144)

General description

Human germinal center–associated lymphoma (HGAL) protein is specifically expressed in the cytoplasm of germinal center B-cells, but is absent in mantle and marginal zone B-cells and in the interfollicular and paracortical regions in normal tonsils and lymph nodes. Its high degree of specificity for germinal center B-cells makes anti-HGAL an ideal marker for the detection of germinal center-derived B-cell lymphomas. Anti-HGAL has the highest overall sensitivity of detecting follicular lymphoma (FL) and in detecting the interfollicular and diffuse components of FL compared with antibodies against bcl2, LMO2, CD10, and bcl6. The addition of anti-HGAL to the immunohistologic panel is beneficial in the work-up of nodal and extranodal B-cell lymphomas, and the efficacy of anti-HGAL in detecting the follicular, interfollicular, and diffuse components of FL is of particular value in the setting of variant immunoarchitectural patterns.

Quality


IVD

IVD

IVD

RUO

Linkage

HGAL Positive Control Slides, Product No. 375S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasodha Natkunam et al.
Blood, 109(1), 298-305 (2006-09-07)
The human germinal-center-associated lymphoma (HGAL) gene and its cognate protein are expressed in a germinal center (GC)-specific manner. Its expression in classic Hodgkin lymphoma (cHL) prompted us to address whether HGAL expression could distinguish biologically distinct subgroups of cHL. Tissue
Russell A Higgins et al.
Archives of pathology & laboratory medicine, 132(3), 441-461 (2008-03-06)
Beginning with the immunologic classifications of Lukes and Collins and Kiel and culminating in the Revised European-American Lymphoma and World Health Organization classifications, the diagnosis of lymphoid tumors relies heavily on the determination of cell lineage, maturation, and function, based
Yasodha Natkunam et al.
Blood, 105(10), 3979-3986 (2005-01-29)
We identified the human germinal center-associated lymphoma (HGAL) in gene-expression profiling studies of diffuse large B-cell lymphoma (DLBCL). The expression of HGAL correlated with survival in patients with DLBCL. The HGAL gene is the human homolog of M17, a mouse
Sheren F Younes et al.
The American journal of surgical pathology, 34(9), 1266-1276 (2010-08-11)
Follicular lymphoma (FL) can exhibit variant histologic patterns that can lead to confusion with other B-cell lymphomas and reactive conditions. Diagnostic markers such as CD10 and BCL2 may be difficult to interpret in variant FL patterns, and are often diminished

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service