Skip to Content
Merck
All Photos(1)

Key Documents

368R-7

Sigma-Aldrich

T-bet (MRQ-46) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-46, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (368R-74)
vial of 0.5 mL concentrate (368R-75)
bottle of 1.0 mL predilute (368R-77)
vial of 1.0 mL concentrate (368R-76)
bottle of 7.0 mL predilute (368R-78)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... TBX21(30009)

General description

T-bet, a T-box transcription factor, is expressed in CD4+ T-lymphocytes committed to T-helper (Th)1 T-cell development from naïve T-helper precursor cells (Thp) and redirects Th2 T-cells to Th1 development. Anti-T-bet is a marker of mature T-cells and is expressed at very low levels in Thp cells and is absent in precursor T-lymphoblastic leukemia/lymphoma cells. Scattered small lymphocytes in the interfollicular T-cell zone of reactive lymphoid tissue, including tonsil, lymph node, and spleen exhibited nuclear staining for anti-T-bet, with no anti-T-bet staining observed in germinal centers or mantle or marginal zones. T-bet is expressed in a significant subset of B-cell lymphoproliferative disorders, particularly at an early stage of B-cell development (precursor B-cell lymphoblastic leukemia/lymphoblastic lymphoma), and B-cell neoplasms derived from mature B-cells, including CLL/SLL, marginal zone lymphoma, and hairy cell leukemia. In contrast, B-cell neoplasms derived from pre-germinal center or germinal center B-cells, including mantle cell lymphoma, follicular lymphoma, diffuse large B-cell lymphoma, and Burkitt lymphoma are negative for anti-T-bet. Therefore, anti-T-bet should serve as a useful marker for the diagnosis and subtyping of B-cell and T-cell lymphoproliferative disorders.

Quality


IVD

IVD

IVD

RUO

Linkage

T-bet Positive Control Slides, Product No. 368S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Dorfman et al.
American journal of clinical pathology, 122(2), 292-297 (2004-08-25)
T-bet, a T-box transcription factor, is expressed in CD4+ T lymphocytes committed to Th1 T-cell development and in a subset of T-cell non-Hodgkin lymphomas. Recent evidence indicates that T-bet also is expressed in B lymphocytes and might participate in immunoglobulin
W X Zhang et al.
Genomics, 70(1), 41-48 (2000-11-23)
The T-box is a strongly conserved protein domain, 174 to 186 amino acids in length, that binds DNA. Many genes from many species have been shown to encode T-box domain-containing proteins. Here we report the cloning and characterization of a
Akira Harashima et al.
Leukemia research, 29(7), 841-848 (2005-06-02)
A number of transcription factors (TFs) have been reported that play crucial roles in hematopoiesis. However, only little is known about how these factors are involved in the mechanisms of hematopoietic development and lineage commitment. To investigate the roles of
Korinna Jöhrens et al.
The American journal of surgical pathology, 31(8), 1181-1185 (2007-08-02)
Diagnosis of minimal infiltration by hairy cell leukemia (HCL) cells in bone marrow trephines is challenging as its differentiation from interstitially distributed infiltrates of reactive B cells and small B-cell lymphomas other than HCL is difficult or impossible. The diagnostic
Teresa Marafioti et al.
The American journal of pathology, 162(3), 861-871 (2003-02-25)
In this study we have investigated the expression of three B-cell-associated transcription factors in normal lymphoid tissue and in T-cell neoplasms (three cell lines, and more than 50 biopsy samples). Nuclear OCT-1 immunoreactivity was seen in normal B cells, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service