Skip to Content
Merck
All Photos(1)

Documents

350R-2

Sigma-Aldrich

FOXP1 (EP137) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP137, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (350R-24)
vial of 0.1 mL concentrate Research Use Only (350R-24-RUO)
vial of 0.5 mL concentrate (350R-25)
vial of 1.0 mL concentrate (350R-26)
vial of 1.0 mL concentrate Research Use Only (350R-26-RUO)
vial of 1.0 mL pre-dilute Research Use Only (350R-27-RUO)
vial of 1.0 mL pre-dilute ready-to-use (350R-27)
vial of 7.0 mL pre-dilute ready-to-use (350R-28)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (350R-28-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500 (concentrated)

isotype

IgG

control

lymph node, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... FOXP1(27086)

General description

Diffuse large B-cell lymphoma (DLBCL) most likely represents different clinicopathologic entities, which are difficult to separate using standard technique. Diffuse large B-cell lymphoma (DLBCL) can be divided into the subtypes of germinal center B-cell–like (GCB), activated B-cell–like (ABC), and unclassified DLBCL. The GCB and ABC subtypes have different pathogenetic mechanisms that will impact the development of targeted therapies. Despite the robustness of gene expression profiling (GEP) in subclassifying DLBCL, GEP techniques are not applicable to the routine clinical practice due to the substantial time, technological expertise, and scarce resources required. Therefore, it is beneficial for the translational application of the GEP classification into protein expression by tumor cells to be developed through immunohistochemical (IHC) staining of formalin-fixed, paraffin-embedded tissues. Different approaches using immunophenotypic algorithms with small panels of antibody biomarkers have been developed to translate the robust information from molecular studies into a routine clinical platform, using antibodies against CD10, BCL6, MUM1/IRF4, GCET1, FoxP1, LMO2, and BCL2. Therefore, FoxP1 is useful in subclassification of DLBCL and a high cutoff (80%) for FoxP1 is needed to achieve high specificity for the ABC subtype.

Quality


IVD

IVD

IVD

RUO

Linkage

FoxP1 Positive Control Slides, Product No. 350S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Alizadeh et al.
Nature, 403(6769), 503-511 (2000-02-17)
Diffuse large B-cell lymphoma (DLBCL), the most common subtype of non-Hodgkin's lymphoma, is clinically heterogeneous: 40% of patients respond well to current therapy and have prolonged survival, whereas the remainder succumb to the disease. We proposed that this variability in
Andreas Rosenwald et al.
The New England journal of medicine, 346(25), 1937-1947 (2002-06-21)
The survival of patients with diffuse large-B-cell lymphoma after chemotherapy is influenced by molecular features of the tumors. We used the gene-expression profiles of these lymphomas to develop a molecular predictor of survival. Biopsy samples of diffuse large-B-cell lymphoma from
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue (2008)
J J F Muris et al.
The Journal of pathology, 208(5), 714-723 (2006-01-10)
Clinical outcome in patients with diffuse large B cell lymphomas (DLBCL) is poorly predictable. Expression of proteins related to germinal centre B (GCB) cell or activated B cells (ABC) and expression of apoptosis-regulating proteins Bcl-2 and XIAP have been found
Lluís Colomo et al.
Blood, 101(1), 78-84 (2002-10-24)
To analyze the relationship between immunophenotyping profile and main clinicopathological features and outcome in diffuse large B-cell lymphoma (DLBCL), we studied 128 patients (59 men, 69 women; median age 65 years) consecutively diagnosed with de novo DLBCL in a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service