Skip to Content
Merck
All Photos(1)

Documents

282M-9

Sigma-Aldrich

HMB-45 (HMB-45) Mouse Monoclonal Antibody

clone HMB-45, unconjugated, Cell Marque

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

HMB-45, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (282M-94)
vial of 0.5 mL concentrate (282M-95)
bottle of 1.0 mL predilute (282M-97)
vial of 1.0 mL concentrate (282M-96)
bottle of 7.0 mL predilute (282M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

melanoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-HMB45 is a useful melanoma immunohistochemical marker that reacts with antigenspresent on immature melanosomes. Anti-HMB45 is useful for identifying amelanoticmelanoma from other neoplastic lesions with similar morphology.

Quality


IVD

IVD

IVD

RUO

Linkage

HMB-45 Positive Control Slides, Product No. 282S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Weiss et al.
The Journal of pathology, 150(3), 195-203 (1986-11-01)
The sequence of events in haematogenous metastasis from colonic carcinoma was analysed, using 1541 necropsy reports from 16 centres. The findings are consistent with the cascade hypothesis that metastases develop in discrete steps, first in the liver, next in the
An assessment of a melanoma-specific antibody (HMB-45) and other immunohistochemical markers of malignant in paraffin-embedded tissue.
AS-Y Leong, et al.
Surgical Pathology, 2, 137-145 (1989)
Helene Nortvig Abrahamsen et al.
Cancer, 100(8), 1683-1691 (2004-04-10)
The optimal technique for sentinel lymph node (SN) assessment in patients with melanoma is controversial. Molecular analysis (reverse transcriptase-polymerase chain reaction) detects significantly greater numbers of SNs with suspected micrometastases (up to 71%) than does routine histopathology (approximately 20%). The
M R Wick et al.
Archives of pathology & laboratory medicine, 112(6), 616-620 (1988-06-01)
The efficacy of two new monoclonal antibodies with cell lineage-restricted reactivity (HMB-45 [melanocytes] and anti-synaptophysin [neuroepithelial cells]) was compared with that of "traditional" antibody panels in the delineation of malignant melanoma (MM) of the sinonasal region, nasopharyngeal carcinoma (NPC), and
L Vaggelli et al.
Tumori, 86(4), 346-348 (2000-10-04)
Elective lymph node dissection (ELND) for patients with malignant melanoma is still controversial. A possible alternative could be biopsy of the first tumor draining lymph node, the sentinel node (SN), which can be identified by means of radionuclide techniques. Our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service