Skip to Content
Merck
All Photos(1)

Key Documents

343R-1

Sigma-Aldrich

TTF-1 (EP229) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP229, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (343R-14)
vial of 0.5 mL concentrate (343R-15)
bottle of 1.0 mL predilute (343R-17)
vial of 1.0 mL concentrate (343R-16)
bottle of 7.0 mL predilute (343R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

lung adenocarcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... A1CF(29974)

General description

Anti-TTF-1 (Thyroid Transcription Factor 1) is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the organs rather than thyroid1-2, germ cell tumors3, and malignant mesothelioma.4-5 It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates.6 TTF-1 labeling is also seen in thyroid and thyroid-derived tumors.7 TTF-1 immunostaining is useful in the differentiation between pulmonary and nonpulmonary origin of adenocarcinomas in malignant effusions.8 TTF-1 staining is very reliable in discerning whether a brain metastasis has arisen from a pulmonary or nonpulmonary site, particularly when dealing with adenocarcinomas and large-cell carcinomas.9

Quality


IVD

IVD

IVD

RUO

Linkage

TTF-1 Positive Control Slides, Product No. 343S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Altman et al.
Gastroenterology, 77(6), 1211-1216 (1979-12-01)
A family was studied in which three middle-aged siblings had unexplained cirrhosis and steatosis. Five of nine additional family members had abnormalities of liver function. Liver biopsy in those 5 revealed steatosis in 3, steatosis and fibrosis in 1, and
C Di Loreto et al.
Cancer letters, 124(1), 73-78 (1998-03-21)
TTF-1 is a tissue-specific transcription factor expressed in the epithelial cells of thyroid and lung. This study investigates the immunohistochemical expression of TTF-1 in pleural malignant mesotheliomas (MM) and adenocarcinomas (AC) of the lung, respectively. For this purpose, 33 biopsy
R Katoh et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 13(5), 570-576 (2000-05-29)
Thyroid transcription factor-1 (TTF-1) has been known to regulate the transcriptional activity of thyroid-specific genes. We examined the expression of TTF-1 in non-neoplastic and neoplastic thyroid tissues. By immunohistochemistry, the nuclei of normal and hyperplastic follicular cells strongly reacted with
M Srodon et al.
Human pathology, 33(6), 642-645 (2002-08-02)
Metastatic carcinoma of unknown primary origin is a perplexing but common problem, accounting for up to 10% to 15% of all solid tumors at presentation. Many of these metastases presumably arise from primary lung carcinomas, but the morphologic features and
K Y Jang et al.
Analytical and quantitative cytology and histology, 23(6), 400-404 (2002-01-05)
To estimate the utility of thyroid transcription factor-1 (TTF-1) and the combined cytokeratin 7 (CK7) and 20 (CK20) immunoprofile as a marker for identifying the primary site of metastatic adenocarcinoma in effusions of the serous cavity. Formalin-fixed, paraffin-embedded cell block

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service