Skip to Content
Merck
All Photos(1)

Key Documents

390M-1

Sigma-Aldrich

GATA3 (L50-823) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

L50-823, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (390M-14)
vial of 0.5 mL concentrate (390M-15)
bottle of 1.0 mL predilute (390M-17)
vial of 1.0 mL concentrate (390M-16)
bottle of 7.0 mL predilute (390M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

breast carcinoma, urothelial carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... GATA3(2625)

General description

GATA binding protein 3 or GATA3, is a zinc finger transcription factor and plays an important role in promoting and directing cell proliferation, development, and differentiation in many tissues and cell types. The human GATA3 gene has been mapped to chromosome10p15. GATA3 expression is primarily seen in breast carcinoma and urothelial carcinoma. Anti-GATA3 can also be useful in the identification of unknown primary carcinoma when carcinomas of the breast or bladder are a possibility

Quality


IVD

IVD

IVD

RUO

Linkage

GATA3 Positive Control Slides, Product No. 390S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Joulin et al.
The EMBO journal, 10(7), 1809-1816 (1991-07-01)
The human GATA1, hGATA1 (previously called NF-E1, GF-1 or Eryf-1), a major sequence-specific DNA-binding protein of the erythrocytic lineage, is a member of a zinc-finger family of DNA-binding proteins. We report here the cloning of a human cDNA for a
M C Labastie et al.
Genomics, 21(1), 1-6 (1994-05-01)
GATA3, a member of the GATA family that is abundantly expressed in the T-lymphocyte lineage, is thought to participate in T-cell receptor gene activation through binding to enhancers. To understand GATA3 gene regulation, we cloned the human gene and the
John P T Higgins et al.
The American journal of surgical pathology, 31(5), 673-680 (2007-04-27)
The morphologic distinction between prostate and urothelial carcinoma can be difficult. To identify novel diagnostic markers that may aid in the differential diagnosis of prostate versus urothelial carcinoma, we analyzed expression patterns in prostate and bladder cancer tissues using complementary
Haiyan Liu et al.
American journal of clinical pathology, 138(1), 57-64 (2012-06-19)
GATA3 expression has been reported in urothelial and breast carcinomas; however, the published data on GATA3 expression in tumors from other organs are limited. Immunohistochemical evaluation of GATA3 expression in 1,110 carcinomas and 310 cases of normal tissue using tissue
Huadong Lai et al.
International journal of cancer, 149(12), 2099-2115 (2021-09-05)
Bladder cancer represents a highly heterogeneous disease characterized by distinct histological, molecular and clinical phenotypes, and a detailed analysis of tumor cell invasion and crosstalks within bladder tumor cells has not been determined. Here, we applied droplet-based single-cell RNA sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service