Skip to Content
Merck
All Photos(1)

Documents

27240

Sigma-Aldrich

Chrysoidine G

for microscopy (Bact., Bot., Vit.)

Synonym(s):

4-Phenylazo-m-phenylenediamine monohydrochloride, Basic Orange 2, Chrysoidine Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4 · HCl
CAS Number:
Molecular Weight:
248.71
Colour Index Number:
11270
Beilstein:
3724653
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Bot., Vit.)

form

powder

mp

235 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Nc1ccc(N=Nc2ccccc2)c(N)c1

InChI

1S/C12H12N4.ClH/c13-9-6-7-12(11(14)8-9)16-15-10-4-2-1-3-5-10;/h1-8H,13-14H2;1H

InChI key

MCTQNEBFZMBRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[A bile-chrysoidine-glycerol culture medium and its use in the diagnosis of gram-negative aerobic bacteria, especially Enterobacteriaceae].
K Ziesché et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 31(9), 516-518 (1985-09-01)
Maggots dyed with chrysoidine.
British medical journal (Clinical research ed.), 289(6456), 1451-1452 (1984-11-24)
Rhiannon M David et al.
Environmental toxicology and chemistry, 28(9), 1893-1900 (2009-04-24)
The use of unicellular algae in ecotoxicity testing is well established, particularly regarding whole-organism and population-level end points such as lethality and population growth. Conflicting information exists, however, on the potential for genetic toxicity to be incorporated into the safety
[Hobby and occupation as causes of cancer].
R L Zielhuis
Nederlands tijdschrift voor geneeskunde, 130(22), 994-998 (1986-05-31)
Ardeshir Shokrollahi et al.
Journal of hazardous materials, 160(2-3), 435-440 (2008-04-25)
A cloud point extraction procedure was presented for the preconcentration of copper(II) ion in various samples. After complexation by 4-(phenyl diazenyl) benzene-1,3-diamine (PDBDM) (chrysoidine), copper(II) ions were quantitatively recovered in Triton X-114 after centrifugation. 0.5ml of methanol acidified with 1.0molL(-1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service