Skip to Content
Merck
All Photos(4)

Key Documents

R6626

Sigma-Aldrich

Rhodamine B

≥95% purity (HPLC), powder

Synonym(s):

Basic Violet 10, Brilliant Pink B, Rhodamine O, Tetraethylrhodamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45170
Beilstein:
4091619
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Rhodamine B, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

titration: suitable

color

faint green to very dark green

mp

210-211 (dec.) (lit.)

solubility

H2O: 1 mg/mL

density

0.79 g/cm3

ε (extinction coefficient)

104500-115800 at 542-554 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccccc4C(O)=O)=[N+](/CC)CC)c1

InChI

1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H

InChI key

PYWVYCXTNDRMGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhodamine B is a xanthene dye,[1] which functions as a water tracer fluorescent. It is used as a colorant in textiles and food stuffs. Rhodamine B is associated with eye, skin and respiratory tract irritation.[2] It is used as a biomarker in oral rabies vaccine.[3]

Application

Rhodamine B has been used:
  • to represent fluorescence of tetra methyl rhodamine (TMR)[4]
  • in the preparation of adhesive resin for confocal laser scanning microscopy[5]
  • as a fluorescent dye in intrahemolymph dye dosing and retinal image acquisition[6]
  • to depict brain-specific dye capture[7]
  • for imaging targets[8]

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palmitate mediated diacylglycerol accumulation causes endoplasmic reticulum stress, Plin2 degradation, and cell death in H9C2 cardiomyoblasts
AliAkoumi
Experimental Cell Research (2017)
Novel models for studying the blood-brain and blood-eye barriers in Drosophila
Nag S, et al.
The Blood-Brain and Other Neural Barriers, 357-369 (2011)
John F. Breidenstein
A study of water circulation in Monterey harbor using rhodamine B dye (1965)
Sulfadimethoxine and rhodamine B as oral biomarkers for European badgers (Meles meles)
Southey AK, et al.
Journal of Wildlife Diseases, 38(2), 378-384 (2002)
5 nm Silver Nanoparticles Amplify Clinical Features of Atopic Dermatitis in Mice by Activating Mast Cells
HanGoo Kang
Nano-Micro Letters (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service