Skip to Content
Merck
All Photos(1)

Key Documents

15759

Sigma-Aldrich

Poloxamer 188

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Lutrol F68

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.25

form

solid

mol wt

~7680-9510 g/mol

composition

oxyethylene, 79.9-83.7%

impurities

≤0.25% ethylene glycol and diethylene glycol
≤0.4% total ash
≤1 ppm ethylene oxide
≤1.0% water
≤2 ppm arsenic traces
≤20 ppm heavy metals
≤5 ppm dioxan
≤5 ppm propylene oxide
50-150 ppm butylhydroxytoluene

ign. residue

≤0.30%

color

APHA: ≤120, 50/50 in CH3OH

pH

5.0-7.5 (100 g/L in H2O)
6.0-7.5 (25 g/L in H2O)

CMC

0.48 mM

transition temp

cloud point >100 °C

HLB

>24

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Stabilizing Function of P 188 in Dermal Formulations: Examined for its stabilizing effects on industrial concentrates of dermal hesperidin smartCrystals, P 188 contributed to enhancing the long-term stability of the formulations, relevant for dermal therapeutic interventions (Romero et al., 2015).

Biochem/physiol Actions

Poloxamer 188 is widely used as a thickening agent and gel binder.

Other Notes

The Poloxamer 188 grades are poloxamers from the extensive line of block copolymers.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were
Xijun Zhang et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 31(6), 842-845 (2011-12-17)
This study examined the effect of P85 (a pluronic block copolymer) and microbubble (MB) ultrasound contrast agents under ultrasound irradiation on gene transfection and expression. The pEGFP plasmids that can encode enhanced green fluorescent protein (pEGFP) served as a report
A N Sklifas et al.
Biofizika, 57(2), 317-324 (2012-05-19)
The adsorption abilities of the perfluorocarbon emulsion stabilized by Proxanol 268 were investigated in vitro and in vivo. In vitro, the saturation point for the blood plasma proteins was nearly reached after five minutes of incubation of the emulsion with
Houjian Gong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13590-13600 (2012-09-08)
The block polyethers PEO-PPO-ph-PPO-PEO (BPE) and PPO-PEO-ph-PEO-PPO (BEP) are synthesized by anionic polymerization using bisphenol A as initiator. Compared with Pluronic P123, the aggregation behaviors of BPE and BEP at an air/water interface are investigated by the surface tension and
Wanhua Feng et al.
Colloids and surfaces. B, Biointerfaces, 105, 43-50 (2013-01-29)
The interaction between bacteria and surfaces is central to many environmental, industrial and medical applications. Surfactants are commonly used in these applications and can potentially influence the bacterium/surface interaction. The effect of surfactants upon bacterial cell surface thermodynamic properties was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service