Merck
  • Combined pluronic P85- and ultrasound contrast agents-mediated gene transfection to HepG2 cells.
Combined pluronic P85- and ultrasound contrast agents-mediated gene transfection to HepG2 cells.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban (2011-12-17)
Xijun Zhang, Kaiyan Li, Xian Cui, Liangjun Hu, Yunchao Chen
ABSTRACT
This study examined the effect of P85 (a pluronic block copolymer) and microbubble (MB) ultrasound contrast agents under ultrasound irradiation on gene transfection and expression. The pEGFP plasmids that can encode enhanced green fluorescent protein (pEGFP) served as a report gene and were mixed with different concentrations of MB/0.05% (w/v) P85. Then the plasmids were transfected into human hepatoma G2 (HepG2) cells. The HepG2 cells treated with MB/P85 or without treatment were exposed to ultrasound (US parameters: 1 MHz, 1.0 W/cm(2), 20 s, 20% duty cycle). Twenty-four hours later, the transfection efficiency was assessed by fluorescence microscopy and fluorescence activated cell sorting (FACS) analysis. The cell viability was evaluated by Trypan blue exclusion test. The results showed that the gene transfection efficiency in HepG2 cells under ultrasound irradiation was significantly higher than that without ultrasound irradiation. HepG2 cells in the MB or P85 group in the absence of ultrasound expressed less amount of green fluorescent protein. The expression efficiency reached (22.14 ± 3.06)% and the survival rate was as high as (55.73 ± 3.32)% in the 30% MB plus P85 group. It was concluded that MB and P85 in the presence of ultrasound can enhance gene transfection and expression.
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000