Skip to Content
Merck
All Photos(1)

Documents

14340C

SAFC

EX-CELL® 325 PF CHO Serum-Free Medium for CHO Cells

protein free, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use
without L-glutamine

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: 3.45 g/L
L-glutamine: no
NaHCO3: 1.6 g/L
HEPES: 7.5 mM
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL™ 325 PF CHO is a protein-free medium which has been developed for the growth of Chinese Hamster Ovary (CHO) cells. Because it contains no large macromolecules, EX-CELL 325 PF CHO facilitates the isolation and purification of secreted proteins from the cells. CHO cells propagated in EX-CELL 302 serum-free media (Catalog No. 14324C and 14326C) can be transferred directly into this protein-free medium without extensive weaning protocols. This medium is supplied without L-glutamine and does not contain purines or pyrimidines to provide an appropriate medium for specialized CHO cell lines (i.e., Glutamine Synthetase, or the GS System™, and DHFR- selection systems).

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sapan J Patel et al.
Electrophoresis, 38(3-4), 417-428 (2016-11-03)
The mechanisms underlying the aberrant growth and interactions between cells are not understood very well. The pre-B acute lymphoblastic leukemia cells directly obtained from an adult patient grow very poorly or do not grow at all at low density (LD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service