Skip to Content
Merck
All Photos(1)

Documents

04076

Sigma-Aldrich

6-(p-Toluidino)-2-naphthalenesulfonyl chloride

suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

2,6-TNS-Cl, 6-(4-Methylanilino)naphthalene-2-sulfonylchloride, 6-[(4-methylphenyl)amino]-2-naphthalenesulfonylchloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14ClNO2S
CAS Number:
Molecular Weight:
331.82
Beilstein:
6521872
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (HPLC)

solubility

DMF: soluble
acetonitrile: soluble

fluorescence

λex 381 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cc1ccc(Nc2ccc3cc(ccc3c2)S(Cl)(=O)=O)cc1

InChI

1S/C17H14ClNO2S/c1-12-2-6-15(7-3-12)19-16-8-4-14-11-17(22(18,20)21)9-5-13(14)10-16/h2-11,19H,1H3

InChI key

NONZPGMJWHNDKP-UHFFFAOYSA-N

Application

6-(p-Toluidino)-2-naphthalenesulfonyl chloride is a fluorescent, hydrophobic probe utilized for labeling peptides to monitor conformational changes of a molecular assembly by measuring alterations in fluorescence intensity .

Analysis Note

Emmax: none
do not dissolve in DMSO

Other Notes

Probe for determining the polarity in biological matrices

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.R. Lakowicz
Principles of Fluorescence Spectroscopy (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service