Skip to Content
Merck
All Photos(11)

Key Documents

1.06009

Supelco

Methanol

≥99.9% (GC), reagent grade, ACS reagent, for extraction and analytical purposes, EMSURE®, reag. Ph. Eur., reag. ISO

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NB.21
grade:
ACS reagent
for extraction
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent
for extraction

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

for analytical purposes

product line

EMSURE®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

dilution

(for analytical testing)

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application

Methanol can be used as a solvent for the extraction of:
  • Bioactive compounds in bottle gourd using gas chromatography-mass spectrometry.
  • Antioxidants in lotus rhizome through centrifugation.
It can also be used as a reactant in the direct synthesis of dimethyl carbonate from CO2.

Features and Benefits

  • Most extensive specifications worldwide
  • Tested for up to 70 parameters
  • Extraordinary purity
  • Very low limiting values
  • Greater accuracy and control of analyses
  • Optimized for highly critical and demandinganalyses
  • Ideal for method development
  • No interference or contamination due tounknown impurities

Linkage

Discover EMSURE - to be more than sure!

Analysis Note

Evaporation Number: 1.9

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta B Fernandes et al.
Frontiers in cell and developmental biology, 8, 335-335 (2020-06-26)
Recent evidence suggests that neural stem cell (NSC) fate is highly dependent on mitochondrial bioenergetics. Tauroursodeoxycholic acid (TUDCA), an endogenous neuroprotective bile acid and a metabolic regulator, stimulates NSC proliferation and enhances adult NSC pool in vitro and in vivo.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service