Skip to Content
Merck
All Photos(1)

Key Documents

01649

Sigma-Aldrich

FITC–DEAE–Dextran

average mol wt 40,000

Synonym(s):

FITC–DEAE–Dextran 40, Fluorescein isothiocyanate–Diethylaminoethyl–Dextran

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

biological source

synthetic

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 40,000

composition

nitrogen content, 2-5%

extent of labeling

0.001-0.020 mol/mol FITC glucose

fluorescence

λex 493 nm; λem 518 nm in 0.1 M Tris pH 8.0

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service