S4132
Sodium carbonate monohydrate
ReagentPlus®, ≥99.5%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Na2CO3 · H2O
CAS Number:
Molecular Weight:
124.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
powder
Recommended Products
grade
reagent
product line
ReagentPlus®
Assay
≥99.5%
form
powder
SMILES string
O.[Na+].[Na+].[O-]C([O-])=O
InChI
1S/CH2O3.2Na.H2O/c2-1(3)4;;;/h(H2,2,3,4);;;1H2/q;2*+1;/p-2
InChI key
MQRJBSHKWOFOGF-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Buffer component, may be used for the removal of peripheral membrane proteins.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Malgorzata Tartanus et al.
Insects, 12(2) (2021-02-06)
The efficacy of two strains of two Beauveria species (B. bassiana and B. brongniartii), individually or as co-inoculants, to control Melolontha sp. grubs was assessed in two organic strawberry plantations in relation to the environmental conditions, their abundance after soil
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Nadine Schlueter et al.
Archives of oral biology, 57(9), 1176-1182 (2012-05-05)
The eroded organic dentine matrix is remarkably resistant to mechanical impacts. Additional brushing abrasion of eroded dentine has only limited influence on tissue loss. Digestive enzymes (e.g., pepsin, trypsin) that can reach the oral cavity during reflux or vomiting can
Yali Zhang et al.
Journal of hazardous materials, 213-214, 167-174 (2012-02-16)
Cyanide tailing is a kind of solid waste produced in the process of gold extraction from gold ore. In this paper, recovery of iron from cyanide tailings was studied with reduction roasting-water leaching process followed by magnetic separation. After analysis
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service