Skip to Content
Merck
All Photos(4)

Documents

M9272

Sigma-Aldrich

Magnesium chloride hexahydrate

ACS reagent, 99.0-102.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

99.0-102.0%

form

powder or crystals

impurities

≤0.005% Insoluble matter

anion traces

nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Ba: ≤0.005%
Ca: ≤0.01%
Fe: ≤5 ppm
K: ≤0.005%
Mn: ≤5 ppm
NH4+: ≤0.002%
Na: ≤0.005%
Sr: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium chloride hexahydrate (MgCl2·6H2O) is an inorganic salt that can be generated from the acid leach solution of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in solar cookers. The result of altering poly (vinyl alcohol)(PVA) films with MgCl2·6H2O has been investigated. The ability of MgCl2·6H2O to reduce the flammability of cotton fabrics has been studied.

Application

Magnesium chloride hexahydrate has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of poly (vinyl alcohol) films by the addition of magnesium chloride hexahydrate.
Jiang X, et al.
Polym. Eng. Sci., 52(7), 1565-1570 (2012)
Magnesium recovery from magnesite tailings by acid leaching and production of magnesium chloride hexahydrate from leaching solution by evaporation.
Ozdemir M, et al.
International Journal of Mineral Processing, 93(2), 209-212 (2009)
Preparation and Analysis of Crude Autolytic Enzyme Extracts from Staphylococcus aureus.
Vaz F and Filipe SR.
Bio-protocol, 5(24) (2015)
Magnesium Bromide and Magnesium Chloride Hexahydrate Catalyzed One-Pot Synthesis of Dihydropyrimidines via Biginelli Reaction under Solvent-Free Conditions.
Gulten S.
Journal of Heterocyclic Chemistry, 50(2), 391-395 (2013)
Magnesium Chloride Hexahydrate Catalyzed One-Pot Synthesis of 3, 4-Dihydropyrimidin-2-(1 H) ones Under Solvent-Free Conditions.
Zhang GL and Cai XH.
Synthetic Communications, 35(6), 829-833 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service