Skip to Content
Merck
All Photos(1)

Key Documents

72061

Sigma-Aldrich

Sodium triphosphate pentabasic

purum p.a., ≥98.0% (T)

Synonym(s):

Pentasodium tripolyphosphate, Sodium tripolyphosphate pentabasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na5P3O10
CAS Number:
Molecular Weight:
367.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0% (T)
form:
powder
solubility:
water: soluble 148 g/L at 20 °C

grade

purum p.a.

Quality Level

Assay

≥98.0% (T)

form

powder

concentration

57-59% (calc. based on P2O5)

pH

9.0-10.0 (25 °C, 1% in H2O)

solubility

water: soluble 148 g/L at 20 °C

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

InChI

1S/5Na.H5O10P3/c;;;;;1-11(2,3)9-13(7,8)10-12(4,5)6/h;;;;;(H,7,8)(H2,1,2,3)(H2,4,5,6)/q5*+1;/p-5

InChI key

HWGNBUXHKFFFIH-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

for the determination of BSB acc. to DIN 38409-H 51

Other Notes

Straight chain phosphates

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meenakshi Malhotra et al.
International journal of nanomedicine, 8, 2041-2052 (2013-06-01)
Recently, cell-penetrating peptides have been proposed to translocate antibodies, proteins, and other molecules in targeted drug delivery. The proposed study presents the synthesis and characterization of a peptide-based chitosan nanoparticle for small interfering RNA (siRNA) delivery, in-vitro. Specifically, the synthesis
Edith Luginbuehl et al.
BMC microbiology, 11, 4-4 (2011-01-07)
Exopolyphosphatases and pyrophosphatases play important but still incompletely understood roles in energy metabolism, and also in other aspects of cell biology such as osmoregulation or signal transduction. Earlier work has suggested that a human exopolyphosphatase, Prune, might exhibit cyclic nucleotide
Akile Çelik et al.
International journal of biological macromolecules, 89, 406-414 (2016-05-08)
β-Glucosidase was covalently immobilized on chitosan-MWCNTs carrier and its aroma enhancement effect in different tea samples was investigated. Chitosan-MWCNTs carrier was prepared by mixing chitosan with MWCNTs (5:1w/w) and characterization of prepared composite carrier was done by FTIR, TGA and
Emmanuel N Koukaras et al.
Molecular pharmaceutics, 9(10), 2856-2862 (2012-08-01)
This work reports details pertaining to the formation of chitosan nanoparticles that we prepare by the ionic gelation method. The molecular interactions of the ionic cross-linking of chitosan with tripolyphosphate have been investigated and elucidated by means of all-electron density
Jessica L Adams et al.
Journal of acquired immune deficiency syndromes (1999), 62(3), 260-266 (2012-11-01)
The active metabolites of tenofovir (TFV) and emtricitabine (FTC) in peripheral blood mononuclear cells (PBMCs) have been used as markers of long-term antiretroviral (ARV) adherence. However, the process of isolating PBMCs is expensive, complex, and not feasible in many settings.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service