Skip to Content
Merck
All Photos(2)

Key Documents

T5508

Sigma-Aldrich

Trisodium trimetaphosphate

≥95%

Synonym(s):

Sodium cyclotriphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3P3O9
CAS Number:
Molecular Weight:
305.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥95%
form:
powder

Quality Level

Assay

≥95%

form

powder

SMILES string

[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)O1

InChI

1S/3Na.H3O9P3/c;;;1-10(2)7-11(3,4)9-12(5,6)8-10/h;;;(H,1,2)(H,3,4)(H,5,6)/q3*+1;/p-3

InChI key

UGTZMIPZNRIWHX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Development and characterization of crosslinked hyaluronic acid polymeric films for use in coating processes.: This study explores the development and properties of crosslinked hyaluronic acid polymeric films, which are applied in various coating processes. The research focuses on the synthesis, characterization, and potential industrial applications of these films. The incorporation of trisodium trimetaphosphate as a crosslinking agent enhances the mechanical properties and stability of the films, making them suitable for use in pharmaceutical and biomedical applications (Sgorla et al., 2016).


Other Notes

Cyclic phosphates

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meimei Liu et al.
Drug delivery, 14(6), 397-402 (2007-08-19)
Hydrogel systems of konjac glucomannan (KGM) cross-linked with trisodium trimetaphosphate (STMP) were prepared for colon-targeting drug delivery. Swelling degrees of the hydrogels were measured in artificial gastrointestinal fluids and in sodium chloride solution with different concentrations to study their dependence
S Sungur
Annali di chimica, 91(5-6), 313-318 (2001-08-18)
The stability constants of the complexes formed between the rare earth metal ions and the anion of trimetaphosphoric acid have been determined at a temperature of 20 degrees C and an ionic strength of mu = 0.1 by the ion-exchange
Yan Liu et al.
Dental materials : official publication of the Academy of Dental Materials, 27(5), 465-477 (2011-03-01)
This study examined the use of sodium trimetaphosphate (STMP) as a biomimetic analog of matrix phosphoproteins for remineralization of artificial carious-affected dentin. Artificial carious lesions with lesion depths of 300±30μm were created by pH-cycling. 2.5% hydrolyzed STMP was applied to
Iliana Diamanti et al.
Journal of dentistry, 39(9), 619-628 (2011-07-16)
The aim of this in vitro study was to assess the effect of toothpastes containing three different sodium fluoride concentrations and a calcium sodium phosphosilicate system, on root dentine demineralization and remineralization. During a fourteen-day pH-cycling protocol, pre-softened bovine root
M J Moretto et al.
Caries research, 44(2), 135-140 (2010-04-02)
It has been suggested that fluoride products are able to reduce erosive tooth wear. Thus, the purpose of this in vitro study was to evaluate the effect of dentifrices with different fluoride concentrations as well as of a low-fluoridated dentifrice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service