Skip to Content
Merck
All Photos(1)

Key Documents

650560

Sigma-Aldrich

tert-Butyl methyl ether

HPLC Plus, for HPLC, GC, and residue analysis, 99.9%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.07
grade:
HPLC Plus
for residue analysis
Assay:
99.9%
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
55-56 °C (lit.)
vapor pressure:
4.05 psi
Pricing and availability is not currently available.

grade

HPLC Plus
for residue analysis

Quality Level

vapor density

3.1 (vs air)

vapor pressure

4.05 psi

Assay

99.9%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.02% water
<1.0 ppb Fluorescence (quinine) at 365 nm

evapn. residue

≤0.0001%

halogenated residue

≤10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C

density

0.74 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.0
λ: 225 nm Amax: 0.50
λ: 250 nm Amax: 0.10
λ: 300-400 nm Amax: 0.005

application(s)

food and beverages

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl ether is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested.[1] Kinetics of its heat-assisted persulfate oxidation has been investigated at various pH, temperature, oxidant concentration and ionic strength levels. Its biodegradation followed a pseudo-first-order decay model and its pseudo-first-order rate constants were evaluated.[2] MTBE can be synthesized by acid catalyzed reaction between methanol and isobutene.[3] A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method.[4]

Application

Suitable for HPLC, spectrophotometry, environmental testing

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid extraction by methyl-tert-butyl ether for high-throughput lipidomics.
Matyash V, et al.
Journal of Lipid Research, 49(5), 1137-1146 (2008)
Eagleson M.
Concise Encyclopedia Chemistry, 154-154 (1994)
Role of tert-Butyl Methyl Ether (TBME) as an External Donor in Propene Polymerization with Dibutyl Phthalate (DBP)-Containing MgCl2-Supported Ti Catalysts Activated with Al(i-C4H9)3.
Ohnishi R and Konakazawa T.
Macromolecular Chemistry and Physics, 205(14), 1938-1947 (2004)
New approach of catalyst-free biodiesel production from canola oil in supercritical tert-butyl methyl ether (MTBE).
Farobie O, et al.
Fuel: The Science and Technology of Fuel and Energy, 135, 172-181 (2014)
Kun-Chang Huang et al.
Chemosphere, 49(4), 413-420 (2002-10-09)
The kinetics of heat-assisted persulfate oxidation of methyl tert-butyl ether (MTBE) in aqueous solutions at various pH, temperature, oxidant concentration and ionic strength levels was studied. The MTBE degradation was found to follow a pseudo-first-order decay model. The pseudo-first-order rate

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service