Skip to Content
Merck
All Photos(1)

Documents

63087

Sigma-Aldrich

Magnesium nitrate hexahydrate

puriss. p.a., ACS reagent, 98.0-102.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

vapor density

6 (vs air)

Assay

98.0-102.0% (KT)

form

solid

pH

5.0-8.2 (25 °C, 50 mg/mL in H2O)

mp

89 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤1 mg/kg
Ba: ≤10 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤10 mg/kg
Na: ≤10 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium nitrate hexahydrate is a hydrated magnesium salt. A mixture of magnesium nitrate hexahydrate and magnesium chloride hexahydrate has been reported as a potential phase change material (PCM) for the storage of latent thermal energy. It has been employed as a base material in this mixture to evaluate the storage and effective utilization of urban waste heat from emerged co-generation systems. Its crystals belong to the monoclinic crystal system.

Application

Magnesium nitrate hexahydrate may be used as a starting material for the synthesis of Mg-Al spinel precursors, required for the preparation of magnesium aluminate spinel (MgAl2O4) powder.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance of heat charge/discharge of magnesium nitrate hexahydrate and magnesium chloride hexahydrate mixture to a single vertical tube for a latent heat storage system.
Nagano K, et al.
Applied Thermal Engineering, 24(2), 209-220 (2004)
Cell dimensions and space group of magnesium nitrate hexahydrate.
Mozzi RL and Bekebrede WR.
Acta Crystallographica, 14(12), 1296-1297 (1961)
A wet-chemical process yielding reactive magnesium aluminate spinel (MgAl2O4) powder.
Li JG, et al.
Ceramics International, 27(4), 481-489 (2001)
Mohamed Abdellatif Bensegueni et al.
Acta crystallographica. Section C, Structural chemistry, 71(Pt 3), 222-228 (2015-03-04)
Two alkaline earth-tetrazole compounds, namely catena-poly[[[triaquamagnesium(II)]-μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(5)] hemi{bis[μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(2)]bis[triaquamagnesium(II)]} monohydrate], {[Mg(C(2)HN(9))(H(2)O)(3)][Mg(2)(C(2)HN(9))(2)(H(2)O)(6)]0.5·H(2)O}n, (I), and bis[5-(pyrazin-2-yl)tetrazolate] hexaaquamagnesium(II), (C(5)H(3)N(6))[Mg(H(2)O)(6)], (II), have been prepared under hydrothermal conditions. Compound (I) is a mixed dimer-polymer based on magnesium ion centres and can be regarded as the first example
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service