Skip to Content
Merck
All Photos(2)

Key Documents

537675

Sigma-Aldrich

Ethylene glycol monopropyl ether

ReagentPlus®, ≥99.40%

Synonym(s):

2-Propoxyethanol, Propyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1731983
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
150-153 °C (lit.)
vapor pressure:
1.3 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor pressure

1.3 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.40%

form

liquid

dilution

(for general lab use)

impurities

≤0.100% (water)

refractive index

n20/D 1.413 (lit.)

bp

150-153 °C (lit.)

mp

−75 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

SMILES string

CCCOCCO

InChI

1S/C5H12O2/c1-2-4-7-5-3-6/h6H,2-5H2,1H3

InChI key

YEYKMVJDLWJFOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol monopropyl ether (EGPEA) is a straight chain glycol derivative. Toxicity study of EGPEA has been reported.[1] Densities, ultrasonic speeds and various excess molar quantities of binary mixtures of water with ethylene glycol monopropyl ether have been investigated.[2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speeds of sound and excess volumetric properties of mixtures of water with ethylene glycol monopropyl ether at 298.15 K.
Douheret G, et al.
Journal of Molecular Liquids, 80(1), 1-18 (1999)
G V Katz et al.
Environmental health perspectives, 57, 165-175 (1984-08-01)
The acute toxicity of ethylene glycol monopropyl ether (EGPE) and ethylene glycol monopropyl ether acetate (EGPEA) was determined in a series of standardized tests. The oral LD50 in rats was 3089 and 9456 mg/kg EGPE and EGPEA, respectively. Skin irritation
R Loch-Caruso et al.
Environmental health perspectives, 57, 119-123 (1984-08-01)
Intercellular communication most likely plays a significant coordinating role in morphogenesis. Blockage of a specific type of intercellular communication, that mediated by gap junctions, has been proposed as a mechanism of action of some teratogens. Several glycol ethers have recently
W J Krasavage et al.
Teratology, 32(1), 93-102 (1985-08-01)
In order to determine the potential developmental toxicity of ethylene glycol monopropyl ether (EGPE), pregnant rats were exposed to vapor concentrations of 100, 200, 300, or 400 ppm of the compound for 6 hours per day on days 6-15 of
W J Krasavage et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 15(3), 517-527 (1990-10-01)
To determine the potential developmental toxicity of ethylene glycol monopropyl ether (EGPE), groups of pregnant New Zealand white rabbits were exposed to target concentrations of 0, 125, 250, or 500 ppm EGPE vapors for 6 hr a day on Days

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service