Skip to Content
Merck
All Photos(1)

Documents

53071

Supelco

Ethylene glycol butyl ether

analytical standard

Synonym(s):

Ethylene glycol monobutyl ether, 2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥99.5% (GC)

autoignition temp.

473 °F

shelf life

limited shelf life, expiry date on the label

composition

Water, ≤0.1% (coulometer)

expl. lim.

10.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.419 (lit.)
n20/D 1.420

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol butyl ether can find applications as a surfactant in investigating the process of spreading, shrinking and resting regime on the surface of water. It can also find applications as a cosolvent in water-borne paints.

Application

Ethylene glycol butyl ether may be used as an analytical standard for the determination of the analyte in biological samples, environmental samples, cleaning products, detergents, and organic solvent mixtures by gas chromatography (GC) based techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Pastor-Belda et al.
Journal of separation science, 39(12), 2292-2299 (2016-05-01)
A rapid and simple procedure is reported for the determination of six ethylene glycol ethers in cleaning products and detergents using gas chromatography with mass spectrometry. The analytes were extracted from 2.0 g samples in acetonitrile (3 mL) and the
Gas chromatographic determination of some glycol ethers and glycol ether acetates by FID, O-FID, and MS detection. Preliminary applications in simultaneous monitoring of these chemicals in biological matrices
Giachetti C, et al.
Journal of High Resolution Chromatography, 19(7), 383-391 (1996)
Persistence, degradation, and movement of triclopyr and its ethylene glycol butyl ether ester in a forest soil
Lee CH, et al.
Journal of Agricultural and Food Chemistry, 34(6), 1075-1079 (1986)
Spreading, fingering instability and shrinking of a hydrosoluble surfactant on water
Mollaei S and Darooneh.HA
Experimental Thermal and Fluid Science, 86, 98-10 (2017)
Citric acid as corrosion inhibitor for aluminium pigment
Muller.B
Corrosion Science, 46(1), 159-167 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service