Skip to Content
Merck
All Photos(2)

Key Documents

366919

Sigma-Aldrich

Chloroform

ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.04
grade:
ACS spectrophotometric grade
Assay:
≥99.8%
technique(s):
UV/Vis spectroscopy: suitable
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

grade

ACS spectrophotometric grade

Quality Level

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

contains

0.5-1.0% ethanol as stabilizer

technique(s)

UV/Vis spectroscopy: suitable

impurities

Acetone and aldehyde, passes test
Acid and chloride, passes test
Free chlorine (Cl), passes test
H2SO4, passes test (darkened)
<0.03% water

evapn. residue

<0.001%

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C (with ethanol stabilizer)
1.492 g/mL at 25 °C (lit.)

cation traces

Pb: ≤0.05 ppm

λ

H2O reference

UV absorption

λ: 245 nm Amax: 1.00
λ: 255 nm Amax: 0.25
λ: 260 nm Amax: 0.15
λ: 270 nm Amax: 0.05
λ: 290-400 nm Amax: 0.01

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform is a is a colorless, volatile chlorinated organic solvent whose vapors have narcotic effect. It is susceptible to degradation with time which can be suppressed by adding ethanol as a stabilizer. Since ethanol is added in higher concentration it leads to increase in polarity which may have a potential influence on some of the applications. Spectrophotometric grade chloroform shows low UV absorption which makes it an ideal solvent for spectrophotometry.

Application

Chloroform (spectrophotometric grade) was used in the following processes:
  • Synthesis of colloidal hydrophobic CdSe core nanocrystals (quantum dots).
  • Preparing phospholipid solutions.
  • As a solvent in the photolysis reaction of ruthenium complexes.
It may be used as:
  • Solvent for recrystallization.
  • Reference compound to detect the presence of chloroform in Tobacco smokes.
  • Extractant in solvent extraction process.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systematic studies on the breakdown of p,p'-DDT in tobacco smokes. Presence of methyl chloride, dichloromethane, and chloroform in tobacco smokes.
Chopra NM and Sherman LR.
Analytical Chemistry, 44(6), 1036-1038 (1972)
Kinetics and Mechanism of the Photooxidation of Bis (bipyridine) dichlororuthenium (II) and the Photoreduction of Bis (bipyridine) dichlororuthenium (III) in Chloroform.
Sathiyabalan S and Hoggard PE.
Inorganic Chemistry, 34(18), 4562-4571 (1995)
Engineering of ultra-small diagnostic nanoprobes through oriented conjugation of single-domain antibodies and quantum dots.
Sukhanova A, et al.
Protocol Exchange (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 216-216 (1994)
Electrogenerated chemiluminescent emission from an organized (L-B) monolayer of a Ru(bpy)32+-based surfactant on semiconductor and metal electrodes.
Zhang X and Bard AJ.
The Journal of Physical Chemistry, 92(20), 5566-5569 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service