Skip to Content
Merck
All Photos(4)

Key Documents

1.09621

Supelco

Ethylene glycol

for analysis EMSURE® Reag. Ph Eur,Reag. USP

Synonym(s):

Ethylene glycol, Glycol, 1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-473-3
NACRES:
NA.05
Assay:
≥99.5% (GC)
bp:
197.6 °C/1013 hPa
vapor pressure:
0.053 hPa ( 20 °C)
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.
reag. USP

Quality Level

vapor pressure

0.053 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

410 °C

expl. lim.

3.2-15.3 % (v/v)

impurities

≤0.00002% Chloride (Cl)
≤0.0003% Substances reducing potassium permanganate (as O)
≤0.001% Acidity (as CH3COOH)
≤0.005% Formaldehyde

color

APHA: ≤10

refractive index

n20/D 1.431-1.433

pH

6-7.5 (20 °C, 100 g/L in H2O)

bp

197.6 °C/1013 hPa

mp

-14--10 °C

transition temp

flash point 111 °C

density

1.11 g/cm3 at 20 °C

cation traces

Fe: ≤0.00005%

storage temp.

2-30°C

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application

Ethylene Glycol is used as a solvent :
  • in diode laser-based detection of single molecules in solutions.[1]
  • and reducing agent in the synthesis of gold nanoparticle.[2]
Additionally, ethylene glycol is also used an important precursor to polyester fibers and resins.

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Acidity (as CH₃COOH): ≤ 0.001 %
Density (d 20 °C/20 °C): 1.113 - 1.115
Density (d 25 °C/25 °C): 1.10 - 1.12
Refractive index (n 20/D): 1.431 - 1.433
Melting point: -14 - -10 °C
Boiling point: 196.5 - 198.5 °C
Chloride (Cl): ≤ 0.00002 %
Fe (Iron): ≤ 0.00005 %
Formaldehyde: ≤ 0.005 %
Substances reducing potassium permanganate (as O): ≤ 0.0003 %
Readily carbonizable substances: conforms
Sulfated ash: ≤ 0.005 %
Water: ≤ 0.1 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Room Temperature Surfactant-Free Synthesis of Gold Nanoparticles in Alkaline Ethylene Glycol
Ditte R R, et al.
Advanced Chemistry, 5, 900-911 (2023)
Diode laser based detection of single molecules in solutions
M S et al.
Chemical Physics Letters, 254, 223-228 (1996)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service