Skip to Content
Merck
All Photos(1)

Key Documents

244252

Sigma-Aldrich

Perchloric acid

ACS reagent, 70%

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor pressure

6.8 mmHg ( 25 °C)

Assay

70%

form

liquid

reaction suitability

reagent type: oxidant

technique(s)

toxicology assay: suitable

impurities

≤0.001% N compounds
≤5 ppm Silicate and phosphate (as SIO2)

ign. residue

≤0.003%

color

APHA: ≤10

density

1.664 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Fe: ≤1 ppm
heavy metals: ≤1 ppm (by ICP)

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perchloric acid (HClO4), an oxygen acid of chlorine, is strongly acidic in nature. Hydrates of perchloric acid are obtained on its dissolution in water.

Application

Perchloric acid has been used to deactivate chloramine-T during the spectrophotometric determination of collagen content of the rat tibialis anterior (TA) tendons and tendon constructs.

It also may be used in the following studies:
  • Determination of reduced glutathione (GSH) and oxidized glutathione (GSSG) content in liver tissues by high performance liquid chromatography (HPLC).
  • As an oxidizing agent in the estimation of sulfur (S) in plant samples.
  • As a solvent for the dissolution of plutonium.

Packaging

poly-coated bottles available: product numbers 244252-2.5L-CB

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 849-849 (1994)
E Driehuis et al.
PloS one, 15(5), e0231588-e0231588 (2020-05-19)
We have recently established a protocol to grow wildtype human oral mucosa organoids. These three-dimensional structures can be maintained in culture long-term, do not require immortalization, and recapitulate the multilayered composition of the epithelial lining of the oral mucosa. Here
Sarah Calve et al.
Journal of applied physiology (Bethesda, Md. : 1985), 108(4), 875-881 (2010-01-30)
Tissue-engineered tendons, derived from an autologous cell source, have the potential to provide an ideal replacement graft that is biologically compatible and has the ability to adapt to the specific mechanical requirements of the in vivo environment. Scaffold-free tendon constructs
Sulfur in plant materials by digestion with nitric and perchloric acid.
Blanchar RW, et al.
Soil Science Society of America Journal. Soil Science Society of America, 29(1), 71-72 (1965)
Eagleson M.
Concise Encyclopedia Chemistry, 790-790 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service