Skip to Content
Merck
All Photos(2)

Key Documents

236101

Sigma-Aldrich

Barium fluoride

powder, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaF2
CAS Number:
Molecular Weight:
175.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Assay

98%

form

powder

bp

2260 °C (lit.)

density

4.89 g/mL at 25 °C (lit.)

SMILES string

F[Ba]F

InChI

1S/Ba.2FH/h;2*1H/q+2;;/p-2

InChI key

OYLGJCQECKOTOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Primary glass modifier in fluorozirconate glasses.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Large barium fluoride detectors.
Wisshak K and Kappeler F.
Nuclear Instruments & Methods in Physics Research. Section A, Accelerators, Spectrometers, Detectors and Associated Equipment, 227(1), 91-96 (1984)
Synthesis and fluorescence of neodymium-doped barium fluoride nanoparticles.
Bender CM, et al.
Chemistry of Materials, 12(7), 1969-1976 (2000)
Refractive properties of barium fluoride.
Mayakova MN, et al.
Journal of the Optical Society of America, 54(5), 628-630 (1964)
S Tavernier et al.
Medical progress through technology, 17(3-4), 237-241 (1991-01-01)
The work presented is part of a design study for a Positron Emission Tomograph (PET) scanner based on the use of BaF2 scintillator and photosensitive wire chambers. The detection efficiency for gamma radiation of 511 keV is found close to
Jian Ruan et al.
Optics express, 17(7), 5163-5169 (2009-04-01)
Bi-doped BaF(2) crystal was grown by the temperature gradient technique and its spectral properties were investigated. The absorption, emission and excitation spectra were measured at room temperature. Two broadband emissions centered at 1070 and 1500 nm were observed in Bi-doped

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service