Skip to Content
Merck
All Photos(2)

Key Documents

449717

Sigma-Aldrich

Calcium fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Fluorite, Fluorspar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaF2
CAS Number:
Molecular Weight:
78.07
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050110
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

bp

2500 °C (lit.)

density

3.18 g/mL at 25 °C (lit.)

SMILES string

F[Ca]F

InChI

1S/Ca.2FH/h;2*1H/q+2;;/p-2

InChI key

WUKWITHWXAAZEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Calcium fluoride based multifunctional nanoparticles for multimodal imaging - This study explores the integration of ions in the calcium fluoride lattice for enhanced multimodal imaging applications (M Straßer et al., 2017).

  • Preparation and characterization of calcium fluoride nano particles for dental applications - Discusses the preparation of nano-sized calcium fluoride for use in dental composites, aiming to improve dental fillings (MS Al-Ajely et al., 2018).

  • Optimization of Calcium Fluoride Crystallization Process for Treatment of High-Concentration Fluoride-Containing Semiconductor Industry Wastewater - Examines the optimization of calcium fluoride crystallization for treating industrial wastewater (A Sinharoy et al., 2024).

  • Synergetic strengthening mechanism of ultrasound combined with calcium fluoride towards vanadium extraction from low-grade vanadium-bearing shale - Investigates the effects of combining ultrasound and calcium fluoride on the extraction of vanadium from shale (B Chen et al., 2021).

  • Sulfate Doping Promotes Agglomeration of Calcium Fluoride Crystals - Studies the influence of sulfate doping on the agglomeration of calcium fluoride crystals, important for industrial applications (T Wang et al., 2024).


Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Australian Journal of Chemistry, 49, 897-897 (1996)
Bioceram. Proc. Int. Symp. Ceram. Med., 7, 279-279 (1994)
G Rølla
Acta odontologica Scandinavica, 46(6), 341-345 (1988-12-01)
The literature concerning the formation and stability of CaF2 in the oral environment is reviewed. In early work the CaF2 formed during topical application with fluoride was assumed to be beneficial. It was suggested that it could protect the enamel
J M ten Cate
European journal of oral sciences, 105(5 Pt 2), 461-465 (1997-12-12)
Low concentrations of fluoride have a beneficial effect on enamel and dentin de- and remineralization. After fluoride treatments, such as topical applications, rinses or dentifrices, salivary fluoride concentrations decrease exponentially in a biphasic manner to very low concentrations within a
Adchara Padermshoke et al.
Applied spectroscopy, 66(6), 711-718 (2012-06-27)
A novel SiO(2)-deposited CaF(2) (SiO(2)/CaF(2)) substrate for measuring vibrational sum-frequency generation (SFG) spectra of silane-based chemisorbed monolayers in aqueous media has been developed. The substrate is suitable for silanization and transparent over a broad range of the infrared (IR) probe.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service