Skip to Content
Merck
All Photos(2)

Key Documents

217581

Sigma-Aldrich

Barium nitrate

ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(NO3)2
CAS Number:
Molecular Weight:
261.34
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99%
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Assay

≥99%

form

solid

impurities

≤0.01% insolubles

pH

5.0-8.0 (25 °C, 5%)

mp

592 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Ca: ≤0.05%
Fe: ≤2 ppm
K: ≤0.005%
Na: ≤0.005%
Sr: ≤0.1%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

IWOUKMZUPDVPGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Barium nitrate can be used as:
  • A catalyst to synthesize 1,4-dihydropyridines.
  • A promoter precursor in ammonia synthesis.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition of bulk and supported barium nitrate.
Bardwell CJ, et al.
Thermochimica Acta, 613, 94-99 (2015)
Facile Synthesis and Antibacterial, Antitubercular, and Anticancer Activities of Novel 1, 4-Dihydropyridines
Sirisha K, et al.
Arch. Pharm. (Weinheim), 343, 342-352 (2010)
The working state of the barium promoter in ammonia synthesis over an active-carbon-supported ruthenium catalyst using barium nitrate as the promoter precursor
Zeng H, et al.
J. Catal., 211, 33-41 (2002)
[Suicidal poisoning with barium nitrate].
Z Kołaciński
Polski tygodnik lekarski (Warsaw, Poland : 1960), 42(11), 339-341 (1987-03-16)
M Monaco et al.
La Medicina del lavoro, 82(5), 439-445 (1991-09-01)
Barium nitrate, which is used in industry in the production of green signal lights, to remove gases from vacuum tubes, and in the production of barium oxide, was assayed to assess the possible mutagenic effects using both the Ames test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service