Skip to Content
Merck
All Photos(4)

Key Documents

307718

Sigma-Aldrich

Iron(III) sulfate hydrate

97%

Synonym(s):

Ferric sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: iron

SMILES string

[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

RUTXIHLAWFEWGM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Convenient, efficient catalyst for the preparation of aromatic esters from the corresponding acids and alcohols.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, G-S.
Synthetic Communications, 29, 607-607 (1999)
Ferric sulfate hydrate-catalyzed O-glycosylation using glycals with or without microwave irradiation.
Zhang G, et al.
Tetrahedron, 64(2), 339-344 (2008)
Rajendra P Shukla et al.
Talanta, 209, 120560-120560 (2020-01-02)
Antipsychotic clozapine is the most effective medication currently available for schizophrenia. However, clozapine is dramatically underutilized due to its harsh side effects that are not effectively monitored. By continuously monitoring clozapine blood levels, such as use of an implantable glucometer
Carmen Stavarache et al.
Ultrasonics sonochemistry, 9(6), 291-296 (2002-10-31)
The influence of ultrasounds (200 kHz frequency) on the decomposition of chlorobenzene (CB) in a water solution (around 100 ppm concentration) containing iron or palladium sulfates was investigated. The intermediates of the sonolysis were identified, thus allowing a deeper insight
Remi Cazelles et al.
Biomedicines, 9(1) (2020-12-31)
Hydroxyurea, an oral medication with important clinical benefits in the treatment of sickle cell anemia, can be accurately determined in plasma with a transition metal dichalcogenide-based electrochemical sensor. We used a two-dimensional molybdenum sulfide material (MoS2) selectively electrodeposited on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service