Skip to Content
Merck
All Photos(2)

Key Documents

210250

Sigma-Aldrich

Hydroxylamine sulfate

99%

Synonym(s):

Hydroxylammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2OH)2 · H2SO4
CAS Number:
Molecular Weight:
164.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
crystals

grade

for analytical purposes

Assay

99%

form

crystals

mp

170 °C (dec.) (lit.)

SMILES string

NO.NO.OS(O)(=O)=O

InChI

1S/2H3NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H2;(H2,1,2,3,4)

InChI key

VRXOQUOGDYKXFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanone oxime undergoes hydrolytic reaction with sulfuric acid to yield hydroxylamine sulfate. It reacts with sulfur dioxide to afford sulfamic acid.

Application

Hydroxylamine sulfate may be used in the following:
  • Along with NaCl for reduction during the atomic absorption spectrophotometric quantification of mercury at ppb (parts-per-billion) levels in solution.
  • As one of the constituents during the preparation of a coloring solution, impregnated in a cellulose based monitoring tape for the sensitive detection of formaldehyde gas.
  • Simultaneous estimation of hydrarine and hydroxylamine in mixtures by titration with alkaline ferricyanide in presence of zinc sulfate.
Powerful reducing agent. Converts aldehydes and ketones to oximes, and acid chlorides to hydroxamic acids. Used as catalyst, swelling agent, and copolymerization inhibitor in polymerization processes.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and efficient approach for preparation of hydroxylamine sulfate from the acid-catalyzed hydrolysis reaction of cyclohexanone oxime.
Zhao, Fangfang, et al.
Chemical Engineering Journal, 272, 102-107 (2015)
The Preparation of Sulfamic Acid by the Hydroxylamine--Sulfur Dioxide Reaction.
Sisler HH and Audrieth LF.
Journal of the American Chemical Society, 61(12), 3389-3391 (1939)
Determination of submicrogram quantities of mercury by atomic absorption spectrophotometry.
Hatch WR and Ott WL.
Analytical Chemistry, 40(14), 2085-2087 (1968)
Marzhan Sypabekova et al.
Biosensors & bioelectronics, 165, 112365-112365 (2020-07-31)
A miniature biosensing platform based on MgO-based nanoparticle doped optical fiber was developed for the biomolecule detection. The technology used a single mode fiber with MgO-based nanoparticles doped core. The detection was based on collecting the Rayleigh backscattering signatures with
Development of a monitoring tape for formaldehyde using hydroxylamine sulfate and methyl yellow.
NAKANO N, et al.
Analytical Sciences, 10(4), 641-645 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service